Product Image
Invitrogen™

pPIC9K Pichia Vector

The pPIC9K vector carries the kanamycin resistance gene that confers resistance to Geneticin™ Reagent in Pichia. Spontaneous generation of multipleRead more
Have Questions?
Catalog NumberQuantity
V1752020 μg
Catalog number V17520
Price (USD)
1,312.00
20 µg
Quantity:
20 μg
Price (USD)
1,312.00
20 µg
The pPIC9K vector carries the kanamycin resistance gene that confers resistance to Geneticin™ Reagent in Pichia. Spontaneous generation of multiple insertion events can be identified by resistance to increased levels of Geneticin™ Reagent. Pichia transformants are selected on histidine-deficient medium and screened for their level of resistance to Geneticin™ Reagent. The ability to grow in high concentrations of Geneticin™ indicates that multiple copies of the kanamycin resistance gene and the gene of interest are integrated into the genome. The pPIC9K vector directs secretion of expressed proteins. This vector is included in the Multi-Copy Pichia Expression Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR), Gentamicin (GmR)
Product TypePichia Expression Vector
Quantity20 μg
VectorpPIC
Cloning MethodRestriction Enzyme/MCS
PromoterAOX1
Protein TagUntagged
Unit Size20 µg
Contents & Storage
Store at -20°C.

Frequently asked questions (FAQs)

When selecting for blasticidin-resistant transformants in the X-33 strain using pPIC6/pPIC6α vectors, why do I get large and small colonies on YPD plates containing 300 µg/ml blasticidin?

Generally, large colonies represent transformants containing pPIC6/pPIC6α integrants, while small colonies represent transformants containing pPIC6/pPIC6α non-integrants. These non-integrants have transduced the pPIC6/pPIC6α plasmid, and therefore, exhibit a low level of blasticidin resistance in the initial selection process. Upon subsequent screening, these non-integrant transformants do not retain blasticidin resistance.

When choosing a blasticidin-resistant transformant for your expression studies, we recommend that you pick blasticidin-resistant colonies from the initial transformation plate and streak them on a second YPD plate containing the appropriate concentration of blasticidin. Select transformants that remain blasticidin-resistant for further studies.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My transformation is not working. Do you have any suggestions?

Here are some suggestinos:

- Make sure that you have harvested cells during log-phase growth (OD <1.0 generally).
- If electroporation is being used, see the electroporator manual for suggested conditions. Vary electroporation parameters if necessary.
- Use more DNA.
- Use freshly made competent cells.
- If the LiCl transformation method is being used, try boiling the carrier DNA.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My spheroplasting of Pichia worked twice, but hasn't worked since. The OD of the culture simply does not drop.

Here are some things to consider:

- If the OD of cells that are used is too high, they will not spheroplast. Do not overgrow cells.
- Do not use old cells and make sure that they are in log phase of growth.
- Make sure to mix zymolyase well before using. Zymolyase is more of a suspension than a solution.
- Make the PEG solution fresh each time and check the pH.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

What are the different kinds of media used for culturing Pichia pastoris and S. cerevisiae?

Following are the rich and minimal media used for culturing Pichia pastoris and S. cerevisiae:

Rich Media:
S. cerevisiae and Pichia pastoris
YPD (YEPD): yeast extract, peptone, and dextrose
YPDS: yeast extract, peptone, dextrose, and sorbitol

Pichia pastoris only
BMGY: buffered glycerol-complex medium
BMMY: buffered methanol-complex medium

Minimal Media (also known as drop-out media):
S. cerevisiae
SC (SD): Synthetic complete (YNB, dextrose (or raffinose or galactose), and amino acids)

Pichia pastoris
MGY: minimal glycerol medium
MD: minimal dextrose
MM: minimal methanol
BMGH: buffered minimal glycerol
BMMH: buffered minimal methanol

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Is there a recommended protocol for fermentation using constitutive expression vectors such as pGAPZ?

Use the following high cell density protocol for pGAP clones. Feed carbon until the desired density is reached (300 to 400 g/L wet cell weight (WCW)). If the protein is well-behaved in the fermenter, increase to 300-400 g/L WCW as with methanol inducible clones. These densities can be reached in less than 48 hours of fermentation. We have fermented constitutive expressers on glycerol using these protocols with good results. Some modifications to the Fermentation Basal Salts Medium that you might want to make are:

1) Substitute 2% dextrose for the 4% glycerol in the batch medium.
2) Substitute 40% dextrose for the 50% glycerol in the fed-batch medium.
3) Feed the 40% dextrose at 12 mL/L/hr (Jim Cregg has published data on expression using several carbon sources as substrates; dextrose gave the highest levels of expression).
4) Yeast extract and peptone may be added to the medium for protein stability.

One warning: If you are working with His- strains, they remain His- after transformation with pGAPZ. Fermentation in minimal medium will require addition of histidine to the fermenter.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Citations & References (12)

Citations & References
Abstract
Inhibition of cell migration and angiogenesis by the amino-terminal fragment of 24kD basic fibroblast growth factor.
Authors:Ding L, Donate F, Parry GC, Guan X, Maher P, Levin EG.
Journal:J Biol Chem
PubMed ID:12063256
'The 24-kDa form of basic fibroblast growth factor inhibits the migration of endothelial cells and mammary carcinoma cells while continuing to promote cell proliferation. This molecule consists of the 18-kDa fibroblast growth factor sequence plus an additional 55 amino acids at the amino-terminal end. Antibody neutralization studies suggested that the ... More
Yeast-secreted bovine herpesvirus type 1 glycoprotein D has authentic conformational structure and immunogenicity.
Authors:Zhu X, Wu S, Letchworth GJ 3rd
Journal:Vaccine
PubMed ID:9178470
'Bovine herpesvirus-1 (BHV-1) glycoprotein D (gD), an envelope glycoprotein, engenders mucosal and systemic immunity protecting cattle from viral infection. Production of gD with authentic immunogenicity is required for a subunit vaccine. We placed the truncated BHV-1 gD gene, lacking its putative transmembrane and cytoplasmic domains, under the control of the ... More
Expression of functional mouse 5-HT5A serotonin receptor in the methylotrophic yeast Pichia pastoris: pharmacological characterization and localization.
Authors:Weiss HM, Haase W, Michel H, Reilander H
Journal:FEBS Lett
PubMed ID:8549774
'The methylotrophic yeast Pichia pastoris was tested for heterologous expression of the mouse 5-HT5A receptor. Three different expression plasmids were constructed where the cDNA of the receptor was cloned under the transcriptional control of the highly inducible promoter of the P. pastoris alcohol oxidase 1 (AOX1) gen. The expression plasmids ... More
Facile production of native-like kappa-bungarotoxin in yeast: an enhanced system for the production of a neuronal nicotinic acetylcholine receptor probe.
Authors:Fiordalisi JJ, James PL, Zhang Y, Grant GA
Journal:Toxicon
PubMed ID:8711755
'Research on the mammalian central nervous system had been hindered by the limited number and meager supply of naturally occurring toxins that can be used as pharmacological reagents. The kappa-neurotoxins in particular are not found abundantly in nature and are difficult to obtain and isolate in quantities sufficient for research ... More
Isolation and expression of a cDNA clone encoding an Alternaria alternata Alt a 1 subunit.
Authors:De Vouge MW, Thaker AJ, Curran IH, Zhang L, Muradia G, Rode H, Vijay HM
Journal:Int Arch Allergy Immunol
PubMed ID:8957113
'Alternaria alternata is recognized as an important source of fungal aeroallergens. Alt a 1, the major allergen of this mold, is a dimer of disulfide-linked subunits that migrate in SDS-PAGE under reducing conditions at apparent M(r)s of 14,500 and 16,000. IgE antibodies to this protein are present in the sera ... More