Product Image
Invitrogen™

pBAD/Myc-His Kit

The pBAD/Myc-His Kit provides all of the necessary reagents to express your protein in a tightly regulated fashion. The pBAD/Myc-HisRead more
Have Questions?
Catalog NumberQuantity
V440011 kit
Catalog number V44001
Price (USD)
1,258.00
1 kit
Quantity:
1 kit
Price (USD)
1,258.00
1 kit
The pBAD/Myc-His Kit provides all of the necessary reagents to express your protein in a tightly regulated fashion. The pBAD/Myc-His vector expresses native proteins or fusion proteins with a C-terminal tag. The vector provides:

• The araBAD promoter for tightly regulated expression
• Translation initiation signals optimized for E. coliexpression
• C-terminal polyhistidine (6xHis) tag for purification with nickel-chelating resin or detection with an Anti-His(C-term) Antibody
• C-terminal c-myc epitope for detection and analysis with an Anti-myc Antibody

Three vectors are provided (A, B, and C). Each has the C-terminal tag in a different reading frame relative to the multiple cloning site to simplify in-frame cloning of your gene.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
CleavageEK (Enterokinase) Recognition Site
Constitutive or Inducible SystemInducible
Inducing AgentArabinose
Product TypeBacterial Expression Vector
Quantity1 kit
Selection Agent (Eukaryotic)None
VectorpBAD
Cloning MethodRestriction Enzyme/MCS
PromoteraraBAD
Protein Tagc-Myc Epitope Tag
Unit Size1 kit
Contents & Storage
The pBAD/Myc-His Kit includes 20 μg of pBAD/Myc-His A, B, & C vector, 20 μg of pBAD/Myc-His/lacZ, 20% L-arabinose, LMG194 and TOP10 E. coli stabs. Vectors and 20% L-arabinose should be stored at -20°C. Store LMG194 and TOP10 stabs at 2 - 8°C. All components are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

How much L-arabinose should I use to induce expression with the pBAD expression system?

While the amount of L-arabinose can vary depending on your expression experiment, we suggest performing a pilot expression experiment with varying amounts of L-arabinose from 0.00002% to 0.2%.

Should I use TOP10 cells or the LMG194 E. coli strain you offer for expression with my pBAD system?

Top10

Advantages:
- Saves time, can go directly from cloning to expression.
- The glycerol stock is more stable because these strains are endA- and recA-.

Disdvantages:
- This strain is not protease-deficient. Therefore, the protein may be degraded.

LMG194

Advantages:
- Grows well in minimal media, except M9.
-Have to transform the plasmid into the cells just for expression.
-RM medium with glucose to ensure low basal level of protein.

Disadvantages:
- Not protease-deficient. Therefore, the protein may be degraded.
- The glycerol stock may not be stable because this cell strain is not recA- or endA-.

What competent cells do you recommend I use for expression with my pBAD expression system?

We recommend using a competent cell strain that is araBADC- and araEFGH+, allowing transportation of L-arabinose, but not metabolizing it. This is important for expression studies, as the level of L-arabinose will be constant inside the cell and will not decrease over time. We offer our TOP10 competent cells, or our LMG194 E. coli strain.

Can you tell me the sequence of primers that can be used in pBAD vectors to sequence my gene of interest?

The pBAD vectors contain a forward and reverse pBAD primer flanking the gene of interest. The sequences are as follows:

pBAD forward primer: 5'-ATGCCATAGCATTTTTATCC-3'

pBAD reverse primer: 5'-GATTTAATCTGTATCAGG-3'

Do you have a list of the cap colors for the pBAD vectors?

Here are the cap colors:

- pBAD/His A: Red

- pBAD/His B: Orange

- pBAD/His C: Yellow

- pBAD/His LacZ: Green

- pBad/gIII A: Yellow

- pBad/gIII B: Green

- pBad/gIII C: Blue

- pBAD/gIII/calmodulin: Purple

Citations & References (6)

Citations & References
Abstract
The intercellular signaling activity of the Mycobacterium tuberculosis chaperonin 60.1 protein resides in the equatorial domain.
Authors:Tormay P, Coates AR, Henderson B,
Journal:J Biol Chem
PubMed ID:15677470
'The major heat shock protein, chaperonin 60, has been established to have intercellular signaling activity in addition to its established protein-folding function. Mycobacterium tuberculosis is one of a small proportion of bacteria to encode two chaperonin 60 proteins. We have demonstrated that chaperonin 60.1 from this bacterium is a very ... More
Novel pathway for arsenic detoxification in the legume symbiont Sinorhizobium meliloti.
Authors:Yang HC, Cheng J, Finan TM, Rosen BP, Bhattacharjee H,
Journal:J Bacteriol
PubMed ID:16199569
'We report a novel pathway for arsenic detoxification in the legume symbiont Sinorhizobium meliloti. Although a majority of ars operons consist of three genes, arsR (transcriptional regulator), arsB [As(OH)3/H+ antiporter], and arsC (arsenate reductase), the S. meliloti ars operon includes an aquaglyceroporin (aqpS) in place of arsB. The presence of ... More
CopA: An Escherichia coli Cu(I)-translocating P-type ATPase
Authors:Rensing C, Fan B, Sharma R, Mitra B, Rosen BP
Journal:Proc Natl Acad Sci U S A
PubMed ID:10639134
'The copA gene product, a putative copper-translocating P-type ATPase, has been shown to be involved in copper resistance in Escherichia coli. The copA gene was disrupted by insertion of a kanamycin gene through homologous recombination. The mutant strain was more sensitive to copper salts but not to salts of other ... More
The prodrug activator EtaA from Mycobacterium tuberculosis is a Baeyer-Villiger monooxygenase.
Authors:Fraaije MW, Kamerbeek NM, Heidekamp AJ, Fortin R, Janssen DB,
Journal:J Biol Chem
PubMed ID:14610090
'EtaA is a newly identified FAD-containing monooxygenase that is responsible for activation of several thioamide prodrugs in Mycobacterium tuberculosis. It was found that purified EtaA displays a remarkably low activity with the antitubercular prodrug ethionamide. Hinted by the presence of a Baeyer-Villiger monooxygenase sequence motif in the EtaA sequence, we ... More
Genetic engineering of phytochrome biosynthesis in bacteria.
Authors: Gambetta G A; Lagarias J C;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11553807
The bilin prosthetic groups of the phytochrome photoreceptors and the light-harvesting phycobiliprotein antennae arise from the oxygen-dependent ring opening of heme. Two ferredoxin-dependent enzymes contribute to this conversion: a heme oxygenase and a bilin reductase with discrete double-bond specificity. Using a dual plasmid system, one expressing a truncated cyanobacterial apophytochrome ... More