Can a Vanquish Charged Aerosol Detector run outside a Vanquish stack and hook into other systems?
Absolutely. It can be integrated or used as an external detector; choose digital or analog output kits for third-party connectivity. Control/traceability are supported via Thermo Scientific Chromeleon CDS.
Can we switch between charged aerosol detection (CAD) and another detector without replumbing?
Yes. The integrated diverter valve lets you direct flow to CAD, bypass to waste, or route to another detector, improving robustness and simplifying multidetector methods.
Do we need a gas supply to use a charged aerosol detector? What are the requirements?
Use nitrogen (recommended) or clean compressed air at about 70–80 psi with typical consumption &ler;4 L/min; turnkey nitrogen generators with integrated air compressors are available to keep supply stable.
Which Vanquish Charged Aerosol Detector model should I choose, FP or HP?
Vanquish Charged Aerosol Detector FP simplifies operation with preset evaporation temperatures and a single acquisition channel; HP adds fine evaporation temperature control (10–100 °C in 0.1 K), multichannel acquisition of up to four power values for linear calibration range optimization, higher data rates and the option for standalone installation with digital or analog control for maximum flexibility. Choose the model with the most important attributes for your needs.
Why would I pick a Vanquish Charged Aerosol Detector (CAD) over UV or other options?
Thermo Scientific Vanquish Charged Aerosol Detector (CAD) offers near-universal detection with a uniform response for nonvolatile and many semivolatile analytes, enabling accurate quantitation across a wide dynamic range—even when chromophores are absent.
Is the Vanquish Flex UHPLC System flow path biocompatible and easy to plumb?
Yes. The Vanquish Flex UHPLC System is fully biocompatible and uses Thermo Scientific Viper Fingertight Fitting Systems across the flow path, delivering near-zero dead volume and tool-free setup.