Product Image
Invitrogen™
WesternBreeze™ Chromogenic Kit, anti-goat
WesternBreeze® Chromogenic Kits detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directlyRead more
Have Questions?
Catalog NumberQuantity
WB71071 kit
Catalog number WB7107
Price (USD)
526.65
Online exclusive
560.00
Save 33.35
1 kit
Quantity:
1 kit
Price (USD)
526.65
Online exclusive
560.00
Save 33.35
1 kit
WesternBreeze® Chromogenic Kits detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots). Detection is accomplished with a ready-to-use BCIP/NBT substrate for alkaline phosphatase. Non-fading purple precipitates develop at the protein bands on the membrane. WesternBreeze® chromogenic offers:
• Clear background
• High sensitivity-low picogram levels detectable
• High specificity
• One simple protocol-no optimization required
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cross ReactivityGoat
Quantity1 kit
ReactivityGoat
Shipping ConditionWet Ice
Substrate TypeAP (Alkaline Phosphatase) Substrate
Detection MethodChromogenic
Membrane CompatibilityNitrocellulose, PVDF
Product LineWesternBreeze
Product TypeWestern Blot Kit
Unit Size1 kit
Contents & Storage
The WesternBreeze™ Chromogenic Kits include blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chromogenic substrate, wash solutions, and incubation trays. Each kit contains complete reagents for 20 mini-blots. Store the kits at +4°C. Guaranteed stable for 6 months when properly stored.
Frequently asked questions (FAQs)
Why is the actual band size on a western blot different from the predicted size of the protein?
Western blotting is based on the separation of proteins by their size on a gel. However, migration of proteins through the gel matrix is also affected by other factors, which may cause the observed band size to be different from the predicted size.
Common causes are:
-Post-translational modification; for example phosphorylation and glycosylation increase the size of the protein
-Post-translation cleavage; many proteins are synthesized as precursor proteins, and then cleaved to give the active form
-Multimers, for example dimerization of a protein. This is usually prevented under reducing conditions, although strong interactions can result in the appearance of higher bands
-Splice variants; alternative splicing may result in different sized proteins being produced from the same gene
-Relative charge; the composition of amino acids (charged vs. non-charged)
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the standard lysis buffers used with mammalian cells for detection of protein expression by immunoprecipitation (IP) or Western blot analysis?
The most commonly used buffer is RIPA Buffer with SDS. We offer RIPA Buffer (Cat. Nos. 89900 and 89901). We also offer the Pierce IP Lysis buffer (Cat. Nos. 87787 and 87788) as well as M-PER (Cat. Nos. 78501, 78503, and 78505).
Find additional tips, troubleshooting help, and resources within our Cell Lysis and Fractionation Support Center.
What conditions do you recommend for overnight Western transfer?
Doing an overnight Westerm transfer is not the preferred method but can be done. The power should be lowered and the buffer should be chilled or the unit should be placed in the cold room to prevent overheating. You may try an overnight transfer at 5-15 V and adjust accordingly. You may also wish to put a second membrane behind the first in order to bind any proteins that transfer through the first membrane. You can use both membranes for staining, immunoblotting, or analysis.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
After transferring onto a PVDF membrane, what is the best way to store the membrane for future probing?
We recommend air drying the PVDF membrane and placing it in an envelope, preferably on top of a supported surface to keep the membrane flat. It can be stored indefinitely at ≤80 degrees C. Right before probing, we recommend re-wetting the membrane with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with the blocking step.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Which transfer buffers are recommended for peptide (N-terminal) sequencing?
Use non-glycine based buffers such as the NuPAGE Invitrogen Transfer buffer, CAPS, or 1/2X TBE transfer buffer.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.