Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product Type | Concentrate System |
Description | Concentrate System - A complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens. |
Includes | 120 Filtering Devices, 120 Conical Centrifuge Tubes, 120 Screw Caps, 30 mL Surfactant |
Quantity | 120/Pk. |
Volume (English) | 1.69 oz. |
Volume (Metric) | 50 mL |
Unit Size | Each |
Product Type | Concentrate System |
Description | Concentrate System - A complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens. |
Includes | 50 Filtering Devices, 50 Disposable 15mL Centrifuge Tubes and Caps, Reagent A (Surfactant), Reagent B (Ethyl Acetate) |
Quantity | 50/Pk. |
Volume (English) | 0.51 oz. |
Volume (Metric) | 15 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Volume (English) | Volume (Metric) | Price (ARS) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
23-005-50 | Each | 1.69 oz. | 50 mL | Contact Us › | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
23-005-51 | Each | 0.51 oz. | 15 mL | Contact Us › | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
50mL Concentrate System:
15mL Concentrate System:
Note:
All PROTOCOL™ Parasitology products are mercury free.