| 75009627 | Full specifications | Each | Thermo Scientific SL4R Plus-MD, 220V 60Hz | 220 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4R Plus-MD, 220V 60Hz | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | Yes, CFC-Free, Pre-Temp Available | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Temperature Range | -10°C to +40°C, ±2°C | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 746 x 361 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 29.4 in. | Width (Metric) Exterior | 746 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 361 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 120 kg | Electrical Requirements | 220 V 60 Hz | Voltage | 220 V | Wattage | 1950 w | Frequency | 60 Hz | Unit Size | Each |
|
| 75009512 | Full specifications | Each | Thermo Scientific SL4 Plus-MD, 208-240V 50/60 Hz | 208/240 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4 Plus-MD, 208-240V 50/60 Hz | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | No | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 566 x 362 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 22.3 in. | Width (Metric) Exterior | 566 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 362 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 89 kg | Electrical Requirements | 208/240 V 50/60 Hz | Voltage | 208/240 V | Wattage | 1600 w | Frequency | 50/60 Hz | Unit Size | Each |
|
| 75009513 | Full specifications | Each | Thermo Scientific SL4 Plus-MD, 120V | 120 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4 Plus-MD, 120V | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | No | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 566 x 362 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 22.3 in. | Width (Metric) Exterior | 566 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 362 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 89 kg | Electrical Requirements | 120 V | Voltage | 120 V | Wattage | 1300 w | Frequency | 50/60 Hz | Unit Size | Each |
|
| 75009527 | Full specifications | Each | Thermo Scientific SL4R Plus-MD, 220V-240V 50Hz/230V 60Hz | 220/240 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4R Plus-MD, 220V-240V 50Hz/230V 60Hz | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | Yes, CFC-Free, Pre-Temp Available | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Temperature Range | -10°C to +40°C, ±2°C | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 746 x 361 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 29.4 in. | Width (Metric) Exterior | 746 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 361 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 120 kg | Electrical Requirements | 220/240 V 50 Hz, 230 V 60 Hz | Voltage | 220/240 V | Wattage | 1950 w | Frequency | 50/60 Hz | Unit Size | Each |
|
| 75009528 | Full specifications | Each | Thermo Scientific SL4R Plus-MD, 120V | 120 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4R Plus-MD, 120V | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | Yes, CFC-Free, Pre-Temp Available | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Temperature Range | -10°C to +40°C, ±2°C | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 746 x 361 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 29.4 in. | Width (Metric) Exterior | 746 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 361 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 120 kg | Electrical Requirements | 120 V | Voltage | 120 V | Wattage | 1450 w | Frequency | 60 Hz | Unit Size | Each |
|
| 75009827 | Full specifications | Each | Thermo Scientific SL4R Plus, 220V 60Hz | 220 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4R Plus, 220V 60Hz | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | No | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Temperature Range | -10°C to +40°C, ±2°C | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 746 x 361 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 29.4 in. | Width (Metric) Exterior | 746 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 361 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 120 kg | Electrical Requirements | 220 V 60 Hz | Voltage | 220 V | Wattage | 1950 w | Frequency | 60 Hz | Unit Size | Each |
|
| 75009912 | Full specifications | Each | Thermo Scientific SL4 Plus, 208-240V 50/60 Hz | 208/240 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4 Plus, 208-240V 50/60 Hz | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | No | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 566 x 362 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 22.3 in. | Width (Metric) Exterior | 566 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 362 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 89 kg | Electrical Requirements | 208/240 V 50/60 Hz | Voltage | 208/240 V | Wattage | 1600 w | Frequency | 50/60 Hz | Unit Size | Each |
|
| 75009927 | Full specifications | Each | Thermo Scientific SL4R Plus, 220V-240V 50Hz/230V 60Hz | 220/240 V | No | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4R Plus, 220V-240V 50Hz/230V 60Hz | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 25,830 x g Fixed Angle (with 30 x 2 mL Rotor), 5,590 x g Swing-Out (with BIOShield 1000A Rotor) | Refrigerated | Yes, CFC-Free, Pre-Temp Available | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Energy efficient, less waste | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | Material | Front, top and sides are out of plastic(PC-ABS) painted with 2K lacquer(PUR based), metal housing is out of cold rolled steel with powder paint(structured), centrifuge bowl is corrosion-resistant high thermal conductivity(STAINLESS STEEL 1.4301 COLD ROLLED). The unit feet are out of rubber. | Model | SL4 Plus | Noise Level | <64 dB(A) Rotor F15-8x50c; 14500 RPM | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | 13 Rotor Options Available, rotor sold separately | Rotor Included | No | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Temperature Range | -10°C to +40°C, ±2°C | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | 5 Year Limited | Dimensions (L x W x H) Exterior | 690 x 746 x 361 mm | Length (English) Exterior | 27.2 in. | Length (Metric) Exterior | 690 mm | Width (English) Exterior | 29.4 in. | Width (Metric) Exterior | 746 mm | Height (English) Exterior | 14.2 in. | Height (English) Exterior Lid Open | 33.9 in. | Height (Metric) Exterior | 361 mm | Height (Metric) Exterior Lid Open | 861 mm | Depth (English) | 27.2 in. | Depth (Metric) | 690 mm | Net Weight | 120 kg | Electrical Requirements | 220/240 V 50 Hz, 230 V 60 Hz | Voltage | 220/240 V | Wattage | 1950 w | Frequency | 50/60 Hz | Unit Size | Each |
|
| 75016075 | Full specifications | Each | Thermo Scientific SL4R Plus, 220V-240V 50Hz/230V 60Hz, TX-1000 Cell Culture Package | 220/240 V | Yes | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4R Plus, 220V-240V 50Hz/230V 60Hz, TX-1000 Cell Culture Package | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Refrigerated | Yes, CFC-Free, Pre-Temp Available | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Less waste, Energy efficient | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | Material | See individual centrifuge, rotor and accessory data | Model | SL4 Plus | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | Cell Culture Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 15mL and 50mL Conical Tube Adapters | Rotor Included | Yes | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Temperature Range | -10°C to +40°C, ±2°C | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | Centrifuge: 5 Year Limited, Rotor: See Manual | Net Weight | See individual components of package | Electrical Requirements | 220/240 V 50 Hz, 230 V 60 Hz | Voltage | 220/240 V | Frequency | 50/60 Hz | Unit Size | Each |
|
| 75016076 | Full specifications | Each | Thermo Scientific SL4 Plus 220V-240V 50Hz/230V 60Hz, TX-1000 Cell Culture Package | 208/240 V | Yes | Contact Us › |
Certifications/Compliance | EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15 | Controller Type | Microprocessor | Description | Thermo Scientific SL4 Plus 220V-240V 50Hz/230V 60Hz, TX-1000 Cell Culture Package | Capacity | 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 rotors) | Max. RCF | 3,374 x g | Max. Speed | 3,800 rpm | Refrigerated | No | Display | Bright LCD Push-Button User Interface | Drive System | Direct, Brushless Induction Low Profile | For Use With (Equipment) | Purchase of Thermo Scientific rotor and accessories required for operation | Green Features | Less waste, Energy efficient | Includes | centrifuge include following in packaging: printed english user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity | Material | See individual centrifuge, rotor and accessory data | Model | SL4 Plus | Profile (Acceleration/Braking) | Choices of 9 Accel/10 Decel Rates | Program Storage | Up to 6 Programs via LCD User Interface buttons | Rotor | Cell Culture Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 15mL and 50mL Conical Tube Adapters | Rotor Included | Yes | Run Time | Up to 9 hr. 59 min. | Standards | ISO 14971, ISO 13485, ISO 9001 | Timer | Option to count up or down | Type | Benchtop Centrifuge | Warranty | Centrifuge: 5 Year Limited, Rotor: See Manual | Net Weight | See individual components of package | Electrical Requirements | 208/240 V 50/60 Hz | Voltage | 208/240 V | Frequency | 50/60 Hz | Unit Size | Each |
|