Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Immunogen sequence: VQTFKGLKLR FEQQRERQDN PKLDSMRSIL RNHRYRRDAR TLEDEEEMWF NTDEDDMEDG EAVVSPSDKT KNDDDIMDPI SKFMERKKLK ESEEKEVLLK TNLSGRQSPS FKLSLSSGTK TNLTSQSSTT NLPGSPGSPG SPGSPGSPGS VPKNTSQTAA ITTKGGLVGL VDYPDDDEDD DEDEDKEDTL PLSKKAKFDS; Positive Samples: HeLa, HepG2, Jurkat, MCF-7, Mouse spleen, Rat brain; Cellular Location: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center
Regulatory subunit of serine/threonine-protein phosphatase 4. May regulate the activity of PPP4C at centrosomal microtubule organizing centers. The PPP4C-PPP4R2-PPP4R3A PP4 complex specifically dephosphorylates H2AFX phosphorylated on 'Ser-140' (gamma-H2AFX) generated during DNA replication and required for DNA DSB repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine/threonine-protein phosphatase 4 regulatory subunit 3A; SMEK homolog 1; SMEK homolog 1, suppressor of mek1; unnamed protein product
Gene Aliases: 1110034C04Rik; BC064465; FLFL1; KIAA2010; mKIAA2010; MSTP033; PP4R3; PP4R3A; PPP4R3A; RGD1309059; SMEK1; smk-1; smk1
UniProt ID: (Human) Q6IN85, (Mouse) Q6P2K6
Entrez Gene ID: (Human) 55671, (Mouse) 68734, (Rat) 314388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support