Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TRPV1 Antibodies

          Alomone Labs, LTD

          TRPV1 (VR1) Polyclonal Antibody

          View all (24) TRPV1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TRPV1 (VR1) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          TRPV1 (VR1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRPV1 (VR1) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRPV1 (VR1) Antibody (ACC-030-200UL) in ICC/IF

          Expression of TRPV1 in rat DRG primary culture - Immunocytochemical staining of paraformaldehyde-fixed and permeabilized rat dorsal root ganglia (DRG) primary cells with Anti-TRPV1 (VR1) Antibody (#ACC-030), (1:200).A. Staining followed by goat Anti-rabbit-AlexaFluor-555 secondary Antibody . B. Nuclear staining using the cell-permeable dye Hoechst 33342. C. Merged image of panels A and B. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRPV1 (VR1) Antibody in Immunocytochemistry (ICC/IF)
          TRPV1 (VR1) Antibody in Immunohistochemistry (IHC)
          TRPV1 (VR1) Antibody in Western Blot (WB)
          TRPV1 (VR1) Antibody in Western Blot (WB)
          TRPV1 (VR1) Polyclonal Antibody

          Product Details

          ACC-030-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)EDAEVFKDSMVPGEK, corresponding to amino acid residues 824-838 of rat TRPV1, Intracellular, C-terminus
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Amount

          160 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).

          Target Information

          TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5' UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Capsaicin receptor; cation channel; DKFZp434K0220; ion channel; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TrpV1; truncated capsaicin receptor protein; Vanilloid receptor; Vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR1; VR1 receptor

          View more View less

          Gene Aliases: OTRPC1; TRPV1; TRPV1_SON; TRPV1alpha; TRPV1beta; VR-1; VR.5'sv; VR1; Vr1l; Vr1l1

          View more View less

          UniProt ID: (Human) Q8NER1, (Rat) O35433, (Mouse) Q704Y3

          View more View less

          Entrez Gene ID: (Human) 7442, (Rat) 83810, (Mouse) 193034

          View more View less

          Function(s)
          nucleotide binding transmembrane signaling receptor activity ion channel activity extracellular ligand-gated ion channel activity excitatory extracellular ligand-gated ion channel activity voltage-gated calcium channel activity calcium channel activity protein binding calmodulin binding ATP binding calcium-release channel activity phosphatidylinositol binding metal ion binding phosphoprotein binding temperature-gated ion channel activity cation channel activity cation transmembrane transporter activity ligand-gated ion channel activity chloride channel regulator activity identical protein binding ion channel
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter temperature homeostasis fever generation diet induced thermogenesis peptide secretion lipid metabolic process ion transport calcium ion transport cell surface receptor signaling pathway chemosensory behavior response to pH response to heat urinary bladder smooth muscle contraction sensory perception of pain calcium-mediated signaling ion transmembrane transport cellular response to heat response to pain behavioral response to pain sensory perception of taste sensory perception of mechanical stimulus thermoception detection of temperature stimulus involved in thermoception detection of temperature stimulus involved in sensory perception of pain detection of chemical stimulus involved in sensory perception of pain protein homotetramerization transmembrane transport excitatory postsynaptic potential smooth muscle contraction involved in micturition calcium ion transmembrane transport cellular response to alkaloid cellular response to ATP cellular response to acidic pH calcium ion import across plasma membrane response to capsazepine calcium ion import into cytosol microglial cell activation inflammatory response signal transduction positive regulation of cytosolic calcium ion concentration response to organonitrogen compound glutamate secretion positive regulation of apoptotic process response to peptide hormone positive regulation of nitric oxide biosynthetic process positive regulation of gastric acid secretion cellular response to cytokine stimulus cellular response to tumor necrosis factor cellular response to growth factor stimulus cellular response to temperature stimulus negative regulation of establishment of blood-brain barrier cellular response to nerve growth factor stimulus transport cation transport catagen cation transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline