Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • GFP Antibodies

          Invitrogen

          GFP Recombinant Rabbit Monoclonal Antibody

          Advanced Verification
          82 Published Figures
          267 References
          View all (134) GFP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GFP Recombinant Rabbit Monoclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (82)
          • Advanced Verification (1)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GFP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 85

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GFP Antibody (G10362) in ICC/IF

          Immunofluorescent analysis of GFP Tag was performed using H3-GFP construct transfected in HEK-293E cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with GFP Recombinant Rabbit Monoclonal Antibody (Product # G10362) at 1:100 dilution and Histone H3 Monoclonal Antibody (865R2) Product # AHO1432) at 1:200 dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Goat ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GFP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GFP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          GFP Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Western Blot (WB)
          GFP Antibody in Immunoprecipitation (IP)
          GFP Antibody in Immunoprecipitation (IP)
          GFP Antibody in Immunoprecipitation (IP)
          GFP Antibody in Immunoprecipitation (IP)
          GFP Antibody in ChIP Assay (ChIP)
          GFP Antibody

          Product Details

          G10362

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:20-1:200
          View 41 publications 41 publications

          Immunohistochemistry (IHC)

          1:20-1:200
          View 115 publications 115 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 5 publications 5 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 7 publications 7 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          1:100
          View 45 publications 45 publications

          Flow Cytometry (Flow)

          Assay-dependent
          View 3 publications 3 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 31 publications 31 publications

          ChIP assay (ChIP)

          -
          View 3 publications 3 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Immunomicroscopy (IM)

          -
          View 2 publications 2 publications

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Affinity Purification (AP)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 8 publications 8 publications
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Pig, Tag

          Host/Isotype

          Rabbit / IgG

          Expression System

          Expi293

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Immunogen

          Full-length GFP

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Ambient

          RRID

          AB_2536526

          Product Specific Information

          Storage and reconstitution: to prepare a 0.2 mg/mL stock solution, reconstitute the lyophilized antibody in 0.5 mL PBS, pH 7.4. Reconstituted product can be stored for up to 3 months at 4°C with the addition of 2mM sodium azide.

          Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

          Target Information

          The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: GFP; GFP tag; GFP2; green fluorescence; green fluorescent; green fluorescent protein; Turbo eGFP

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline