Concanavalin A, Alexa Fluor™ 350 Conjugate, 5 mg - FAQs
Concanavalin A, Alexa Fluor™ 350 Conjugate, 5 mg - FAQs
View additional product information for Concanavalin A (Con A) Conjugates - FAQs (C11254, C860, C11252, C11253, C825, C21402, C56126, C56127, C21421, C21401)
This product does not have any FAQs available.