Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ZO-2 Antibodies

          Invitrogen

          ZO-2 Polyclonal Antibody

          Advanced Verification
          82 Published Figures
          138 References
          View all (14) ZO-2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZO-2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (82)
          • Advanced Verification (2)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 87

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZO-2 Antibody (71-1400) in ICC/IF

          Immunofluorescent analysis of ZO2/TJP2 Antibody was done on 90% confluent log phase CaCo2 cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with ZO2/TJP2 Antibody (Product # 71-1400) at 1 µg/mL in 1% BSA and incubated for 3 hours at room temperature and then labeled with Alexa Fluor 488 Goat Anti-Rabbit IgG Secondary Antibody (Product # A-11008) at a dilution of 1:400 for 45 minutes at roo... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry, Immunoprecipitation (ICC/IF, IP)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry, Immunoprecipitation (WB, ICC/IF, IP)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          ZO-2 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunocytochemistry (ICC/IF)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Western Blot, Immunohistochemistry, Immunoprecipitation (WB, IHC, IP)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Immunohistochemistry (IHC)
          ZO-2 Antibody in Western Blot, Immunohistochemistry, Immunoprecipitation (WB, IHC, IP)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          ZO-2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          ZO-2 Antibody in Western Blot (WB)
          ZO-2 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          ZO-2 Antibody
          ZO-2 Antibody

          Product Details

          71-1400

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          View 51 publications 51 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 20 publications 20 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 3 publications 3 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 3 publications 3 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1 µg/mL
          View 36 publications 36 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          -
          View 13 publications 13 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          GST Pull Down (GST-PD)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse

          Published species

          Dog, Fish, Human, Mouse, Pig, Plant, Rabbit, Rat, Sheep, Xenopus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide derived from the central portion of the ZO-2 protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533976

          Target Information

          CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Friedreich ataxia region gene X104 (tight junction protein ZO-2); Tight junction protein 2; Tight junction protein ZO-2; Tight junction protein ZO-2 (Zonula occludens 2 protein) (Zona occludens 2 protein) (Tight junction protein 2); unnamed protein product; X104; ZO-2A; ZO-2C; zona occludens 2; Zona occludens protein 2; Zonula occludens protein 2

          View more View less

          Gene Aliases: C9DUPq21.11; DFNA51; DUP9q21.11; FHCA1; PFIC4; TJP2; X104; ZO-2; ZO2

          View more View less

          UniProt ID: (Dog) Q95168, (Human) Q9UDY2, (Mouse) Q9Z0U1

          View more View less

          Entrez Gene ID: (Dog) 403854, (Human) 9414, (Mouse) 21873

          View more View less

          Function(s)
          protein binding guanylate kinase activity protein domain specific binding protein binding, bridging cadherin binding cell adhesion molecule binding protein tyrosine kinase binding protein C-terminus binding tight junction
          Process(es)
          homotypic cell-cell adhesion maintenance of blood-brain barrier cell-cell junction organization GMP metabolic process GDP metabolic process intestinal absorption establishment of endothelial intestinal barrier regulation of membrane permeability cell-cell adhesion protein localization to cell-cell junction positive regulation of maintenance of permeability of blood-brain barrier
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline