Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ANT1/ANT2/ANT3/ANT4 Antibodies

          Invitrogen

          ANT1/ANT2/ANT3/ANT4 Polyclonal Antibody

          View all (2) ANT1/ANT2/ANT3/ANT4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ANT1/ANT2/ANT3/ANT4 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          ANT1/ANT2/ANT3/ANT4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ANT1/ANT2/ANT3/ANT4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ANT1/ANT2/ANT3/ANT4 Antibody (PA5-109394) in IHC (P)

          Immunohistochemistry analysis of ANT1/ANT2/ANT3/ANT4 in paraffin-embedded human colon carcinoma. Samples were incubated with ANT1/ANT2/ANT3/ANT4 Polyclonal antibody (Product # PA5-109394) using a dilution of 1:50 (40x lens). Perform microwave antigen retrieval with 10 mM PBS buffer pH 7.2 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ANT1/ANT2/ANT3/ANT4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANT1/ANT2/ANT3/ANT4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANT1/ANT2/ANT3/ANT4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANT1/ANT2/ANT3/ANT4 Antibody in Western Blot (WB)
          ANT1/ANT2/ANT3/ANT4 Polyclonal Antibody

          Product Details

          PA5-109394

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human ANT1 (NP_001142.2).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.65 mg/mL

          Amount

          165 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2854805

          Product Specific Information

          Immunogen sequence: GMLPDPKNVH IFVSWMIAQS VTAVAGLVSY PFDTVRRRMM MQSGRKGADI MYTGTVDCWR KIAKDEGAKA FFKGAWSNVL RGMGGAFVLV LYDEIKKYV

          Target Information

          ANT1/2/3/4 (SLC25A4, SLC25A5, SLC25A6, SLC25A31) are members of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Adenine nucleotid translocator 2 fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotid translocator 2, fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocase 4; adenine nucleotide translocase-1; Adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); adenine nucleotide translocator 2 fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocator 2, fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 1; ADP,ATP carrier protein 2; ADP,ATP carrier protein 4; ADP,ATP carrier protein, fibroblast isoform; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP.ATP translocase; ADP/ADT translocator protein; ADP/ATP carrier 1; ADP/ATP carrier protein; ADP/ATP translocase 1; ADP/ATP translocase 2; ADP/ATP translocase 4; ANT 1; ANT 2; ANT 4; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; Mitochondrial S-adenosylmethionine carrier protein; mitochondrial S-adenosylmethionine transporter; progressive external ophthalmoplegia 2; progressive external ophthalmoplegia 3; S-adenosylmethionine mitochondrial carrier protein; SAM carrier; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator) member 4 isoform SLC25A4_1; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator) member 4 isoform SLC25A4_2; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator) member 4 isoform SLC25A4_3; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; Solute carrier family 25 member 26; Solute carrier family 25 member 31; Solute carrier family 25 member 4; Solute carrier family 25 member 5; solute carrier family 25, member 26; Sperm flagellar energy carrier protein; SZFULLNAME2; unnamed protein product

          View more View less

          Gene Aliases: 1700034J06Rik; 2F1; 4930433D19Rik; 4933433F13Rik; AAC1; AAC2; AAC4; Anc1; ANT; ANT 1; ANT 4; ANT1; ANT2; ANT4; AU019225; AW557176; COXPD28; D6Bwg0781e; MTDPS12; MTDPS12A; PEO2; PEO3; PEOA2; SAMC; SFEC; SFEC35kDa; SLC25A26; SLC25A31; SLC25A4; SLC25A5; Slc25a6; SZGENESYMBOL2; T1; T2; T3

          View more View less

          UniProt ID: (Human) Q70HW3, (Human) Q9H0C2, (Human) P12235, (Human) P05141, (Mouse) Q5U680, (Mouse) Q3V132, (Rat) Q05962, (Mouse) P48962, (Rat) Q09073, (Mouse) P51881

          View more View less

          Entrez Gene ID: (Human) 115286, (Human) 83447, (Human) 291, (Human) 292, (Mouse) 67582, (Rat) 362403, (Rat) 689108, (Mouse) 73333, (Rat) 85333, (Mouse) 11739, (Rat) 25176, (Mouse) 11740

          View more View less

          Function(s)
          S-adenosyl-L-methionine transmembrane transporter activity antiporter activity S-adenosyl-L-methionine:S-adenosyl-L-homocysteine antiporter activity ATP:ADP antiporter activity protein binding hydrogen ion transmembrane transporter activity adenine transmembrane transporter activity oxidative phosphorylation uncoupler activity adenine nucleotide transmembrane transporter activity RNA binding ubiquitin protein ligase binding transporter activity enzyme binding poly(A) RNA binding mitochondrial carrier protein transfer/carrier protein
          Process(es)
          ion transport S-adenosyl-L-methionine transport purine nucleoside transmembrane transport macromolecule methylation L-alpha-amino acid transmembrane transport mitochondrial S-adenosyl-L-methionine transmembrane transport male meiosis I spermatogenesis cell differentiation regulation of mitochondrial membrane permeability transmembrane transport mitochondrial ADP transmembrane transport negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway mitochondrial ATP transmembrane transport mitochondrial genome maintenance generation of precursor metabolites and energy apoptotic mitochondrial changes adenine transport ADP transport negative regulation of necroptotic process positive regulation of mitophagy hydrogen ion transmembrane transport adaptive thermogenesis chromosome segregation positive regulation of cell proliferation B cell differentiation erythrocyte differentiation adenine nucleotide transport cellular response to leukemia inhibitory factor transport S-adenosyl-L-methionine transmembrane transport liver development heart development aging negative regulation of cardiac muscle cell apoptotic process positive regulation of cell growth involved in cardiac muscle cell development response to interleukin-1 negative regulation of mitochondrial membrane permeability involved in apoptotic process positive regulation of oxidative phosphorylation uncoupler activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline