Search
Search
QED Bioscience
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which may partially inactivate or slightly decrease the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 22.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: big potassium channel beta subunit 3; BK beta 3; BK channel beta subunit 3; BK channel subunit beta-3; BKbeta3; calcium-activated potassium channel regulatory subunit; Calcium-activated potassium channel subfamily M subunit beta-3; Calcium-activated potassium channel subunit beta-3; Calcium-activated potassium channel, subfamily M subunit beta-3; Charybdotoxin receptor subunit beta-3; HBeta3; K(VCA)beta-3; KCNMB3; kcnmb3 {ECO:0000312|EMBL:BAF79924.1}; large conductance, voltage and Ca2+ activated potassium channel Maxi K beta 3 subunit; Maxi K channel subunit beta-3; MaxiK channel beta-subunit 3; potassium channel subfamily M regulatory beta subunit 3; potassium large conductance calcium-activated channel, subfamily M beta member 3; potassium large conductance calcium-activated channel, subfamily M, beta member 4; Slo-beta-3; slobeta3 (KCNMB3); unnamed protein product
Gene Aliases: BKBETA3; EG435726; Gm5707; HBETA3; K(VCA)BETA-3; KCNMB2; KCNMB3; KCNMBL; SLO-BETA-3; SLOBETA3
UniProt ID: (Human) Q9NPA1, (Rat) A7VL23
Entrez Gene ID: (Human) 27094, (Mouse) 100502876, (Rat) 310303
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support