MitoTracker™ Dyes for Mitochondria Labeling
Invitrogen™
MitoTracker™ Dyes for Mitochondria Labeling
The rosamine- and carbocyanine-based staining dyes MitoTracker Orange CMTMRos, Red CMXRos, Red FM, Green FM, and Deep Red FM enable mitochondria visualization with fluorescent imaging.
Have Questions?
更改视图buttonViewtableView
货号数量描述
A6644150 μgMitoTracker™ Green FM
A6644450 μgMitoTracker™ Red FM
A6644250 μgMitoTracker Orange CMTMRos
A6644350 μgMitoTracker™ Red CMXRos
A6644050 μgMitoTracker Deep Red FM
A664675 x 50 μgMitoTracker Deep Red FM
A664685 x 50 μgMitoTracker™ Green FM
A664695 x 50 μgMitoTracker Orange CMTMRos
A664705 x 50 μgMitoTracker™ Red CMXRos
A664725 x 50 μgMitoTracker™ Red FM
货号 A66441
价格(CNY)
589.00
Each
数量:
50 μg
描述:
MitoTracker™ Green FM
价格(CNY)
589.00
Each
The Mitotracker dyes for flow cytometry are cell permeant reagents that label mitochondria in live cells. Provided at an optimized concentration with protocols for flow cytometry to allow for more precise analysis with both conventional and spectral flow cytometry.
The rosamine- and carbocyanine-based staining dyes MitoTracker Orange CMTMRos, Red CMXRos, Red FM, Green FM, and Deep Red FM enable mitochondria visualization with fluorescent imaging. These mitochondria dyes are packaged in vials of 50 μg each to create freshly reconstituted dyes without the impact of freeze-thaw cycles to aid in a simplified sample prep for cell analysis workflow.
Rosamine-based MitoTracker dyes for mitochondria labeling
The cell-permeant MitoTracker probes stain active mitochondria in live cells for labeling and localization in fluorescent cell imaging. Our rosamine-based mitochondrial staining dyes include MitoTracker Orange CMTMRos, a derivative of tetramethylrosamine, and MitoTracker Red CMXRos, a derivative of X-rosamine. The fluorescent signal from the rosamine-based MitoTracker dyes is retained in the mitochondria even after aldehyde fixation and detergent permeabilization, making these mitochondria dyes flexible for many workflows including applications that require subsequent processing such as immunocytochemistry or in situ hybridization. The MitoTracker Red and Orange dyes are well suited for multicolor labeling experiments because their fluorescence is well resolved from the green fluorescence of other probes.
Carbocyanine-based MitoTracker dyes for mitochondria labeling
The cell-permeant MitoTracker dyes stain active mitochondria in live cells for mitochondrial labeling and localization in fluorescent cell imaging. The carbocyanine-based dyes MitoTracker Green FM and MitoTracker Red FM accumulate in active mitochondria but are not well-retained in mitochondria after aldehyde fixation. The carbocyanine-based MitoTracker Deep Red FM stains active mitochondrial in live cells and is well-retained in mitochondria after aldehyde fixation and subsequent permeabilization with detergents for applications that require subsequent processing such as immunocytochemistry or in situ hybridization. The MitoTracker Red FM and MitoTracker Deep Red FM dyes are well suited for multicolor labeling experiments because their red fluorescence is well resolved from the green fluorescence of other probes.
Benefits of using MitoTracker mitochondria labeling dyes
The MitoTracker dyes are provided in vials of 50 μg of lyophilized powder ready for reconstitution. To label mitochondria, live cells are simply incubated with the MitoTracker probe of your choice. The mitochondrial staining dyes passively diffuse across the plasma membrane and accumulate in active mitochondria. The MitoTracker dyes are offered in a range of wavelengths and can be used for mitochondrial localization in multicolor experiments.
Conventional fluorescent stains such as tetramethylrosamine and rhodamine 123 are readily sequestered by active mitochondria and are reversible in dynamic membrane potential measurements as they easily wash out of cells upon loss in membrane potential. In contrast, MitoTracker dyes contain a mildly thiol-reactive chloromethyl moiety so that mitochondrial staining is retained if the mitochondrial membrane potential is lost, allowing many of the MitoTracker dyes to be retained during cell fixation.
Throughout the cell life cycle, mitochondria use oxidizable substrates to produce an electrochemical proton gradient across the mitochondrial membrane (whose potential is negative), resulting in ATP production. The MitoTracker dyes are ideal probes for mitochondria staining in experiments studying the cell cycle or processes such as apoptosis and other end point assays.
仅供研究使用。
规格
颜色Green
描述MitoTracker™ Green FM
检测方法Fluorescence
激发波长范围490/516 nm
适用于(应用)Cellular Imaging, Cell Analysis, Cellular Imaging
适用于(设备)荧光显微镜、流式细胞仪、微孔板读数仪
产品规格Vial
产品线MitoTracker
数量50 μg
运输条件Room Temperature
标签类型Fluorescent Dye
产品类型染料
亚细胞定位Mitochondria/Organelle Stain
Unit SizeEach
内容与储存
Store in freezer (-5 to -30°C), and protect from light.