Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • G3BP1 Antibodies

          Proteintech

          G3BP1 Polyclonal Antibody

          172 References
          View all (36) G3BP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite G3BP1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (24)
          G3BP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          G3BP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          G3BP1 Antibody (13057-2-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed sodium arsenite treated HeLa cells using G3BP1 antibody (Product # 13057-2-AP) at dilution of 1:600 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          G3BP1 Antibody in Immunocytochemistry (ICC/IF)
          G3BP1 Antibody in Immunocytochemistry (ICC/IF)
          G3BP1 Antibody in Immunocytochemistry (ICC/IF)
          G3BP1 Antibody in Immunocytochemistry (ICC/IF)
          G3BP1 Antibody in Immunocytochemistry (ICC/IF)
          G3BP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          G3BP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          G3BP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          G3BP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          G3BP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          G3BP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Western Blot (WB)
          G3BP1 Antibody in Flow Cytometry (Flow)
          G3BP1 Antibody in Flow Cytometry (Flow)
          G3BP1 Antibody in Immunoprecipitation (IP)
          G3BP1 Antibody in Immunoprecipitation (IP)
          G3BP1 Polyclonal Antibody

          Product Details

          13057-2-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          View 58 publications 58 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 88 publications 88 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 14 publications 14 publications

          RNA Immunoprecipitation (RIP)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Chicken, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          G3BP1 Fusion Protein Ag3728 (167-466 aa encoded by BC006997)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.65 mg/mL

          Amount

          97.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: PDDSGTFYD QAVVSNDMEE HLEEPVAEPE PDPEPEPEQE PVSEIQEEKP EPVLEETAPE DAQKSSSPAP ADIAQTVQED LRTFSWASVT SKNLPPSGAV PVTGIPPHVV KVPASQPRPE SKPESQIPPQ RPQRDQRVRE QRINIPPQRG PRPIREAGEQ GDIEPRRMVR HPDSHQLFIG NLPHEVDKSE LKDFFQSYGN VVELRINSGG KLPNFGFVVF DDSEPVQKVL SNRPIMFRGE VRLNVEEKKT RAAREGDRRD NRLRGPGGPR GGLGGGMRGP PRGGMVQKPG FGVGRGLAPR Q (167-466 aa encoded by BC006997)

          Target Information

          This gene encodes one of the DNA-unwinding enzymes which prefers partially unwound 3'-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion. This enzyme is a member of the heterogeneous nuclear RNA-binding proteins and is also an element of the Ras signal transduction pathway. It binds specifically to the Ras-GTPase-activating protein by associating with its SH3 domain. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATP-dependent DNA helicase VIII; DNA helicase VIII; G3BP; G3BP-1; GAP binding protein; GAP SH3 binding protein; GAP SH3 domain-binding protein 1; GTPase activating protein (SH3 domain) binding protein 1; hDH VIII; HDH-VIII; MGC111040; Ras GTPase-activating protein-binding protein 1; Ras-GTPase-activating protein SH3-domain binding protein 1; Ras-GTPase-activating protein SH3-domain-binding protein; RasGAP-associated endoribonuclease G3BP; unnamed protein product

          View more View less

          Gene Aliases: AI849976; B430204O07; C87777; G3BP; G3BP1; HDH-VIII; mKIAA4115; RGD1310666

          View more View less

          UniProt ID: (Human) Q13283, (Mouse) P97855

          View more View less

          Entrez Gene ID: (Human) 10146, (Rat) 171092, (Mouse) 27041

          View more View less

          Function(s)
          nucleotide binding nucleic acid binding DNA binding DNA helicase activity RNA binding RNA helicase activity mRNA binding helicase activity nuclease activity endonuclease activity protein binding ATP binding hydrolase activity ATPase activity DNA/RNA helicase activity ribosomal small subunit binding molecular condensate scaffold activity poly(A) RNA binding RNA metabolism protein nucleic acid metabolism protein
          Process(es)
          immune system process Ras protein signal transduction positive regulation of type I interferon production stress granule assembly innate immune response defense response to virus positive regulation of stress granule assembly negative regulation of canonical Wnt signaling pathway transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline