Thermo Scientific Chemicals

Adenosine-5'-diphosphate trilithium salt, 98%

Catalog number: J64497.14
25 g, Each
Thermo Scientific Chemicals

Adenosine-5'-diphosphate trilithium salt, 98%

Catalog number: J64497.14
25 g, Each
Quantity
Catalog number: J64497.14
also known as J64497-14
Price (EUR)
Price: 193,00
Online price: 173,70
Your price:
Quantity
-

Description

It is used as a salt formulation of the essential metabolic compound adenosine diphosphate (ADP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a salt formulation of the essential metabolic compound adenosine diphosphate (ADP).

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Not Hazardous

    Not Hazardous