Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FLMTCENSSV VGINTSYEGI PQDLYVENDI FADCNVDCNC PSKIWDPVCG NNGLSYLSAC LAGCETSIGT GINMVFQNCS CIQTSGNSSA VLGLCDKGPD CSLMLQYF
The organic anion transporting polypeptides, OATP-A (also designated OATP1, OATP1A2 and SLC21A3) and OATP-C (also designated OATP2, SLC21A6 and LST1), mediate hepatic uptake of cardiac glycosides. The expression of OATP-C, but not OATP-A, is inducible by phenobarbital and pregnenolone-16a-carbonitrile, resulting in the increased capacity of the liver to extract cardiac glycosides from the plasma. OATP-A is expressed in liver and kidney and helps mediate sodium-independent uptake of the anionic steroid conjugates dehydroepiandrosterone sulfate, estradiol-17-glucuronide and prostaglandin. OATP-C is exclusively expressed in liver and is localized to the basolateral hepatocyte membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain digoxin carrier protein; Brain-specific organic anion transporter; OATP-1; OATP-A; OATP-B1; OATP1A2; organic anion transporter 2; organic anion transporting polypeptide A; Organic anion-transporting polypeptide 1; SO1A2; Sodium-independent organic anion transporter; Sodium-independent organic anion transporter 1; Sodium-independent organic anion-transporting polypeptide 1; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family (organic anion transporter) member 1; solute carrier family (organic anion transporter) member 3; solute carrier family 21 (organic anion transporter), member 1; solute carrier family 21 (organic anion transporter), member 3; solute carrier family 21 (organic anion transporter), member 5; Solute carrier family 21 member 1; Solute carrier family 21 member 3; Solute carrier family 21 member 5; solute carrier family 21, member 1; Solute carrier organic anion transporter family member 1A1; Solute carrier organic anion transporter family member 1A2; Solute carrier organic anion transporter family member 1A4; solute carrier organic anion transporter family, member 1a4; unnamed protein product
Gene Aliases: A530084B21; OATP; OATP-1; OATP-A; OATP1; Oatp1a1; OATP1A2; Oatp1a4; Oatp2; Slc21a1; SLC21A3; Slc21a5; Slco1a1; SLCO1A2; Slco1a4
UniProt ID: (Human) P46721, (Mouse) Q9QXZ6, (Rat) P46720, (Rat) O35913
Entrez Gene ID: (Human) 6579, (Mouse) 28248, (Rat) 50572, (Rat) 170698
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support