Thermo Scientific Chemicals

Tétrafluoroborate d’ammonium, 97 %, Thermo Scientific Chemicals

Référence: 011488.36
500 g, Each
Thermo Scientific Chemicals

Tétrafluoroborate d’ammonium, 97 %, Thermo Scientific Chemicals

Référence: 011488.36
500 g, Each
Quantité
Référence: 011488.36
également connu sous le numéro 011488-36
Prix (EUR)
Prix 55,80
Prix en ligne: 50,22
Votre prix
Quantité
-

Identifiants chimiques

CAS
13826-83-0
IUPAC Name
ammonium tetrafluoroboranuide
Molecular Formula
BF4H4N
InChI Key
PDTKOBRZPAIMRD-UHFFFAOYSA-O
SMILES
[NH4+].F[B-](F)(F)F
Assay (unspecified)
96.1% min

Description

Ammonium fluoroborate is used as an analytical reagent. In the textile printing and dyeing industry, it used as a catalyst for resin finishing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician