Thermo Scientific™
Poudre de graphite, naturelle, microcristalline, APS 2-15 micron, 99,9995 % (base en métaux), Thermo Scientific Chemicals
Référence: 014736.TC
227 g, Each
Thermo Scientific™
Poudre de graphite, naturelle, microcristalline, APS 2-15 micron, 99,9995 % (base en métaux), Thermo Scientific Chemicals
Référence: 014736.TC
227 g, Each
Quantité
Total Metal Impurities
0.0005% max.
Description
Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Figures
Documentation et téléchargements
Certificats
    Foire aux questions (FAQ)
    Citations et références
    Search citations by name, author, journal title or abstract text
    Sécurité et manipulation
    Not Hazardous
    Not Hazardous