Thermo Scientific Chemicals

Lingot de fer, 99,95 % (base de métaux), Thermo Scientific Chemicals

Référence: 042385.18
50 g, Each
Thermo Scientific Chemicals

Lingot de fer, 99,95 % (base de métaux), Thermo Scientific Chemicals

Référence: 042385.18
50 g, Each
Quantité
Référence: 042385.18
également connu sous le numéro 042385-18
Prix (EUR)
Prix 186,00
Prix en ligne: 165,54
Votre prix
Quantité
-

Identifiants chimiques

CAS
7439-89-6
IUPAC Name
iron
Molecular Formula
Fe
InChI Key
XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES
[Fe]
Appearance (Color)
Silvery
Form
Slug, 3.175mm (0.125in) diameter.
Assay from Supplier's CofA
≥99.95% (metals basis)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    EU Specific Hazard Statements

    May form combustible dust concentrations in air