Thermo Scientific Chemicals

Hexafluorosilicate de potassium, 99,999 % (base de métaux), Thermo Scientific Chemicals

Référence: 044495.18
50 g, Each
Thermo Scientific Chemicals

Hexafluorosilicate de potassium, 99,999 % (base de métaux), Thermo Scientific Chemicals

Référence: 044495.18
50 g, Each
Quantité
Référence: 044495.18
également connu sous le numéro 044495-18
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
16871-90-2
IUPAC Name
dipotassium hexafluorosilanediuide
Molecular Formula
F6K2Si
InChI Key
UUHPPUWEDCVPCO-UHFFFAOYSA-N
SMILES
[K+].[K+].F[Si--](F)(F)(F)(F)F
Total Metal Impurities
0.001% max.

Description

Potassium hexafluorosilicate is used in the production of porceleins. It is also used for the preservation of timber. It is involved in the manufacturing of ceramics, aluminum and magnesium smelting. In addition, it is used as an intermediate in the manufacturing of optical glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute dermal toxicity
    Category 3
    Acute Inhalation Toxicity - Dusts and Mists
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H311 + H331 - Toxic if swallowed, in contact with skin or if inhaled

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P311 - Call a POISON CENTER or doctor/physician