Thermo Scientific Chemicals

Séléniure de cadmium, 99,999 % (base métallique), Thermo Scientific Chemicals

Référence: 045456.18
50 g, Each
Thermo Scientific Chemicals

Séléniure de cadmium, 99,999 % (base métallique), Thermo Scientific Chemicals

Référence: 045456.18
50 g, Each
Quantité
Informational:Informatif:Il est possible que nous ayons besoin d’informations supplémentaires concernant ce produit chimique pour nos rapports de conformité réglementaire et chimique. Nous vous contacterons pour obtenir ces informations une fois votre commande passée, si nécessaire.
Référence: 045456.18
également connu sous le numéro 045456-18
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
1306-24-7
IUPAC Name
selanylidenecadmium
Molecular Formula
CdSe
InChI Key
AQCDIIAORKRFCD-UHFFFAOYSA-N
SMILES
[Se]=[Cd]
Total Metal Impurities
0.001% max.

Description

Cadmium selenide is applied in opto-electronic devices, laser diodes, biomedical imaging, nanosensing, high-efficiency solar cells and thin-film transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO - Usage exclusivement réservé à la recherche

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute Inhalation Toxicity - Dusts and Mists
    Category 3
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H331 - Toxic if swallowed or if inhaled

    H373 - May cause damage to organs through prolonged or repeated exposure

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P260 - Do not breathe dust/fume/gas/mist/vapors/spray

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P311 - Call a POISON CENTER or doctor/physician

    P403 + P233 - Store in a well-ventilated place. Keep container tightly closed