Thermo Scientific Chemicals

N-oxyde de 4-nitropyridine, 97 %, Thermo Scientific Chemicals

Référence: 128921000
100 g, Flacon en verre, Each
Thermo Scientific Chemicals

N-oxyde de 4-nitropyridine, 97 %, Thermo Scientific Chemicals

Référence: 128921000
100 g, Flacon en verre, Each
Quantité
Conditionnement
Référence: 128921000
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
1124-33-0
IUPAC Name
4-nitropyridin-1-ium-1-olate
Molecular Formula
C5H4N2O3
InChI Key
RXKNNAKAVAHBNK-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=[N+]([O-])C=C1
Appearance (Color)
Yellow to brown
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Form)
Powder or crystals
Melting point
159°C to 164°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 - Toxic if swallowed

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing