Thermo Scientific Chemicals

Acétate de cobalt(II) tétrahydraté, + de 98 %, Thermo Scientific Chemicals

Référence: 293060500
50 g, Flacon en plastique, Each
Thermo Scientific Chemicals

Acétate de cobalt(II) tétrahydraté, + de 98 %, Thermo Scientific Chemicals

Référence: 293060500
50 g, Flacon en plastique, Each
Quantité
Conditionnement
Référence: 293060500
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
6147-53-1
IUPAC Name
λ²-cobalt(2+) diacetate tetrahydrate
Molecular Formula
C4H14CoO8
InChI Key
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES
O.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
Appearance (Form)
Crystalline powder or crystals
Nitrogen compounds (as N)
=<0.005 %
Insoluble matter
=<0.02 % (in water)
Copper (Cu)
=<100 ppm
Appearance (Color)
Dark pink to red-pink

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Respiratory Sensitization
    Category 1
    Skin Sensitization
    Category 1
    Germ Cell Mutagenicity
    Category 2
    Reproductive Toxicity
    Category 1B
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H317 - May cause an allergic skin reaction

    H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

    H341 - Suspected of causing genetic defects

    H350i - May cause cancer by inhalation

    H360F - May damage fertility

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P284 - Wear respiratory protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

    Additional EU labelling

    Restricted to professional users