Thermo Scientific Chemicals

2-Bromo-1-indanol, 99 %, Thermo Scientific Chemicals

Référence: A13511.18
50 g, Each
Thermo Scientific Chemicals

2-Bromo-1-indanol, 99 %, Thermo Scientific Chemicals

Référence: A13511.18
50 g, Each
Quantité
Référence: A13511.18
également connu sous le numéro A13511-18
Prix (EUR)
Prix 113,00
Prix en ligne: 101,70
Votre prix
Quantité
-

Identifiants chimiques

CAS
5400-80-6
IUPAC Name
2-bromo-2,3-dihydro-1H-inden-1-ol
Molecular Formula
C9H9BrO
InChI Key
RTESDSDXFLYAKZ-UHFFFAOYNA-N
SMILES
OC1C(Br)CC2=CC=CC=C12
Appearance (Color)
White to pale cream
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
128.0-132.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text