Thermo Scientific Chemicals

gamma-Octanolactone, 98 %, Thermo Scientific Chemicals

Référence: A18142.14
25 g, Each
Thermo Scientific Chemicals

gamma-Octanolactone, 98 %, Thermo Scientific Chemicals

Référence: A18142.14
25 g, Each
Quantité
Référence: A18142.14
également connu sous le numéro A18142-14
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
104-50-7
IUPAC Name
5-butyloxolan-2-one
Molecular Formula
C8H14O2
InChI Key
IPBFYZQJXZJBFQ-UHFFFAOYNA-N
SMILES
CCCCC1CCC(=O)O1
Form
Liquid
Refractive Index
1.4425-1.4465 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text