Thermo Scientific Chemicals

4-Fluoro-2-méthylbenzonitrile, 97 %, Thermo Scientific Chemicals

Référence: H26641.14
25 g, Each
Thermo Scientific Chemicals

4-Fluoro-2-méthylbenzonitrile, 97 %, Thermo Scientific Chemicals

Référence: H26641.14
25 g, Each
Quantité
Référence: H26641.14
également connu sous le numéro H26641-14
Prix (EUR)
Prix 361,00
Prix en ligne: 321,29
Votre prix
Quantité
-

Identifiants chimiques

CAS
147754-12-9
IUPAC Name
4-fluoro-2-methylbenzonitrile
Molecular Formula
C8H6FN
InChI Key
BJBXUIUJKPOZLV-UHFFFAOYSA-N
SMILES
CC1=CC(F)=CC=C1C#N
Melting Point (clear melt)
65.0-75.0?C
Appearance (Color)
White to cream
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or lumps

Description

As organic intermediate, pharmaceutical intermediate. In chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Comme intermédiaire organique, intermédiaire pharmaceutique. En synthèse chimique.

Remarques
Stockez-le dans un endroit frais. Garder le récipient bien fermé dans un endroit sec et bien ventilé. Conserver à l’écart des agents oxydants forts.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water