Thermo Scientific Chemicals

1,4,7,10-tétraazacyclotridécane, Thermo Scientific Chemicals

Référence: H26671.03
1 g, Each
Thermo Scientific Chemicals

1,4,7,10-tétraazacyclotridécane, Thermo Scientific Chemicals

Référence: H26671.03
1 g, Each
Quantité
Référence: H26671.03
également connu sous le numéro H26671-03
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
295-14-7
IUPAC Name
1,4,7,10-tetraazacyclotridecane
Molecular Formula
C9H22N4
InChI Key
LADZJJOUGVGJHM-UHFFFAOYSA-N
SMILES
C1CNCCNCCNCCNC1
Form
Powder or crystalline powder
Proton NMR
Conforms to structure
Carbon-13 NMR
Conforms to structure
Appearance (Color)
White to pale cream
Assay (special)
≥95.0% by proton NMR

Description

1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4,7,10-Le tétraazacylotridecane est utilisé pour trouver les constantes de liaison et pour construire des éléments de bloc.

Solubilité
Soluble dans l’eau.

Remarques
Hygroscopique. Garder le récipient hermétiquement fermé. Stocker dans des conditions fraîches et sèches et dans des récipients bien scellés.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention