Thermo Scientific Chemicals

1,6-Diméthoxynaphtalène, 98+ %, Thermo Scientific Chemicals

Référence: L18129.03
1 g, Each
Thermo Scientific Chemicals

1,6-Diméthoxynaphtalène, 98+ %, Thermo Scientific Chemicals

Référence: L18129.03
1 g, Each
Quantité
Référence: L18129.03
également connu sous le numéro L18129-03
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
3900-49-0
Assay (GC)
≥98.0%
Melting Point (clear melt)
54.0-61.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale yellow to yellow to brown to gray

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text