Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Proteins & Peptides ›
          • IL-13 Proteins

          Gibco

          Human IL-13 Recombinant Protein, PeproTech®

          122 References
          View all (10) IL-13 proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human IL-13 Recombinant Protein, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (2)
          Human IL-13 Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human IL-13 Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human IL-13 Protein (200-13-2UG) in Bioactivity

          Bioassay analysis of Human IL-13 Recombinant Protein, PeproTech® (Product # 200-13-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human IL-13 Protein in Bioactivity (Bioactivity)
          Human IL-13 Protein in Bioactivity (Bioactivity)

          Product Details

          200-13-2UG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 99 publications 99 publications

          Miscellaneous PubMed (Misc)

          -
          View 16 publications 16 publications

          Western blot control (WB Ctrl)

          Assay-dependent
          -

          ELISA standard (ELISA Std)

          -
          View 4 publications 4 publications

          Bioactivity (Bioactivity)

          Assay-dependent
          View 1 publication 1 publication

          Cell differentiation (Cell Diff)

          -
          View 1 publication 1 publication

          Cell stimulation (Cell Stm)

          -
          View 1 publication 1 publication

          Cell induction (Cell Ind)

          -
          - -
          Product Specifications

          Species

          Human

          Published species

          Human, Mouse, Non-human primate, Pig, Rat

          Expression System

          E. coli

          Amino acid sequence

          MSPGPVPPST ALRELIEELV NITQNQKAPL CNGSMVWSIN LTAGMYCAAL ESLINVSGCS AIEKTQRMLS GFCPHKVSAG QFSSLHVRDT KIEVAQFVKD LLLHLKKLFR EGRFN

          Molecular weight

          12.6 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 98% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          The ED50 as determined by the dose-dependent proliferation of TF-1 cells was ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          2 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          Recombinant Human IL-13 is a 12.6 kDa protein consisting of 115 amino acid residues.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ALRH; ALRH antibody; BHR1; il 13; IL-13; ILN; Interleukin; Interleukin-13; Interleukin13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600 antibody

          View more View less

          Gene Aliases: IL-13; IL13; NC30; P600

          View more View less

          UniProt ID: (Human) P35225

          View more View less

          Entrez Gene ID: (Human) 3596

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline