Thermo Scientific™

Luminaris Color Probe qPCR Master Mix, high ROX

產品號碼: K0333
Thermo Scientific™

Luminaris Color Probe qPCR Master Mix, high ROX

產品號碼: K0333
產品號碼
K0333
包裝規格
1250 reactions
價格 (HKD)
價格 9,574.00
您的價格
供應狀況
***
數量
產品號碼
包裝規格
250 reactions
價格 (HKD)
價格 2,281.00
您的價格
供應狀況
***
數量
產品號碼
包裝規格
500 reactions
價格 (HKD)
價格 4,368.00
您的價格
供應狀況
***
數量
產品號碼
包裝規格
5,000 reactions
價格 (HKD)
價格 26,879.00
您的價格
供應狀況
***
數量
產品號碼包裝規格價格 (HKD)供應狀況數量
K03331250 reactions
價格 9,574.00
您的價格
***
K0331250 reactions
價格 2,281.00
您的價格
***
K0332500 reactions
價格 4,368.00
您的價格
***
K03345,000 reactions
價格 26,879.00
您的價格
***
產品概覽
圖表
影片
建議
建議
文件
常見問題
引用資料與參考文獻
其他資訊
建議
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of Luminaris Color Probe qPCR Master Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific Luminaris Color Probe and Luminaris Probe qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR. The master mixes include Thermo Scientific Hot Start Taq DNA polymerase, uracil-DNA glycosylase (UDG) and dNTPs in an optimized PCR buffer. Only the template, primers and probe need to be added.

The Hot Start Taq DNA polymerase, in combination with an optimized buffer, ensures PCR specificity and sensitivity. dUTP and UDG are included in the mix for carry-over contamination control (see Reference 1). The Luminaris Color Probe qPCR Master Mixes are supplemented with an inert blue dye and a separate Yellow Sample Buffer that contains a yellow dye. The qPCR reaction mix containing both components is green.

The use of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes in qPCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. Individual formulations of Luminaris Color Probe and Luminaris Probe qPCR Master Mixes with ROX at different concentrations as passive reference dye are available for most qPCR platforms (see Real-time PCR instrument compatibility chart).

Highlights

• Blue Master Mix and Yellow Sample Buffer for easy pipetting
• UDG in the master mix to prevent carry-over contamination
• Specificity—Hot Start Taq DNA Polymerase and the optimized buffer reduce primer dimers
• Sensitivitity—single copy detection
• Wide linear range—accurate quantification across 9 orders of magnitude
• Reproducibility and convenience— ready-to-use 2X master mix
• Equal performance of blue and colorless master mixes
• Versions with premixed ROX for different real-time platforms
• Standard cycling protocols

Applications

• Gene expression analysis
• siRNA validation
• Genotyping
• Pathogen detection

Includes

Luminaris Color Probe High ROX qPCR Master Mix (2X) includes Hot Start Taq Polymerase, UDG and dNTPs (also dUTP) in an optimized PCR buffer with a blue dye. The master mix contains high concentration of ROX passive reference dye for specific real-time platforms (see Real-Time PCR instrument compatibility chart) and is supplied with 40X Yellow Sample Buffer and nuclease-free water.
For Research Use Only. Not for use in diagnostic procedures.

規格

Description
Color Probe High ROX
Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line
Luminaris
Product Type
qPCR Master Mix
Quantity
1250 x 20 μL Reactions
Sufficient For
1250 Reactions of 20 μL
For Use With (Application)
Gene Expression, Genotyping, siRNA Validation
Concentration
2X (Luminaris Color Probe High ROX qPCR Master Mix), 40X (Yellow Sample Buffer)
No. of Reactions
1,250 Reactions
Sample Type
DNA, dsDNA
Volume
10 x 1.25 mL

內容物與存放

Contains:
• 10 x 1.25 mL Luminaris Color Probe High ROX qPCR Master Mix (2X) containing Hot Start Taq DNA Polymerase, UDG and dNTPs (with dUTP) in an optimized PCR buffer with a blue dye; sufficient for 1250 x 20 μL reactions. The master mix contains high concentration of ROX passive reference dye for specific real-time platforms.
• 3 x 1.25 mL 40X Yellow Sample Buffer
• 10 x 1.25 mL Nuclease-Free Water

Store at -20°C.

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text