Product Image
Invitrogen™

EXPRESS One-Step Superscript™ qRT-PCR Kit, universal

EXPRESS One-Step SuperScript qRT-PCR kits incorporate proven Invitrogen enzymes specifically tailored to deliver superior results on high-throughput, fast-cycling instruments forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
1178101K2500 Reactions
11781200500 Reactions
Catalog number 1178101K
Price (INR)
555,985.00
Each
No. of Reactions:
2500 Reactions
Price (INR)
555,985.00
Each
EXPRESS One-Step SuperScript qRT-PCR kits incorporate proven Invitrogen enzymes specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for your probe-based qPCR experiments. This fast, one-step mix is designed to work with a variety of instruments’ default protocols, including Applied Biosystems and non-Applied Biosystem (see universal version) qPCR platforms.

Features of the EXPRESS One-Step Superscript qRT-PCR Kit include:
Fast-activating, antibody-mediated Platinum Taq DNA Polymerase–complete, rapid activation; provides high sensitivity and reproducibility
The One-Step SuperScript III Reverse Transcriptase module– a unique formulation to ensure high yield and less PCR inhibition
Flexible formats with separate or pre-mixed ROX–designed to work with default instrument protocols on a wide range of high-throughput fast instruments (e.g., AB 7500 Fast, AB 7900 HT Fast, AB StepOne, Roche LightCycler™ 480, Corbett Rotor-Gene™)
Contains heat-labile UDG–significantly reduces carryover contamination risk with excellent activity from room temperature to approximately 40°C. Due to the thermostability of SuperScript III, the cDNA synthesis step is run at 50°C, at which temperature the heat-labile UDG is totally and irreversibly inactivated

EXPRESS kits work with manufacturer’s default protocols on fast instruments and come in a convenient, stable SuperMix format for ultimate convenience. The kits are offered with either pre-mixed ROX or a separate tube of ROX to fit your instrument needs, as well as SYBR Green chemistry options:
EXPRESS One-Step Superscript qRT-PCR Kit, universal (this page)
EXPRESS One-Step Superscript qRT-PCR Kit, with premixed ROX
EXPRESS One-Step SYBR GreenER Kit, with premixed ROX
EXPRESS One-Step SYBR GreenER Kit, universal
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
FormatTube
No. of Reactions2500 Reactions
PolymeraseTaq DNA Polymerase
Product LineSuperScript
Product TypeqRT-PCR Kit
Quantity2,500 reactions
Sample TypeRNA
Sufficient For2500 Reactions
Detection MethodPrimer-probe
PCR Method1-step RT-qPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
The EXPRESS One-Step SuperScript™ qRT-PCR Universal kit contains 3 modules: 2X EXPRESS SuperScript™ Mix (5 × 1 mL), Express qPCR SuperMix Universal (5 × 5 mL), and ROX Reference Dye (5 × 500 μL).

Store at -20°C for long-term storage. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at 4–8°C.

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.