PDPH (3-(2-pyridyldithio)propionyl hydrazide)
Thermo Scientific™

PDPH (3-(2-pyridyldithio)propionyl hydrazide)

Thermo Scientific Pierce PDPH is a short, pyridyldithiol-and-hydrazide crosslinker for reversibly conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, suchRead more
Have Questions?
Catalog NumberQuantity
2230150 mg
Catalog number 22301
Price (INR)
-
Quantity:
50 mg
Thermo Scientific Pierce PDPH is a short, pyridyldithiol-and-hydrazide crosslinker for reversibly conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).

Features of PDPH :

• Reactive groups: pyridyldisulfide and hydrazide
• Reactive toward: sulfhydryl groups and carbonyl (aldehyde) groups
• Short (9.2A), sulfhydryl-to-aldehyde crosslinker with disulfide bond spacer arm (cleavable)
• Pyridyldithiol group results in attachment to sulfhydryls via disulfide bond, which can be cleaved with DTT, TCEP or other reducing agents
• Hydrazide group conjugates to oxidized sugars of glycoproteins and carbohydrates
• Use sodium meta-periodate to oxidize glycosylation (e.g., sialic acid) to reactive aldehyde groups
• Use with EDC to conjugate primary amine of hydrazide group to carboxyl groups

Product References:
Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityYes
DescriptionPDPH
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)229.32
PEGylatedNo
Product LinePierce
Quantity50 mg
Reactive MoietyHydrazide, Pyridyldisulfide
Shipping ConditionAmbient
SolubilityDMF, DMSO
Spacer Arm Length9.2 Å
Water SolubleNo
Chemical ReactivityCarbohydrate-Sulfhydryl
CleavableBy Thiols
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for PDPH (3-(2-pyridyldithio)propionyl hydrazide)?

PDPH (3-(2-pyridyldithio)propionyl hydrazide) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

We are using EDC to crosslink a protein onto carboxyl-modified beads. We would like to add in a linker molecule to extend the radius of beads to increase the surface area so more proteins can be immobilized. Is this possible?

To provide a linker between the beads and the target protein, you would need a molecule with a primary amine (to conjugate to the carboxyl group on the beads with EDC) on one end and another reactive group on the other (to conjugate to the protein). For instance, sulfo-SMCC (Cat. No. A39268) could be used in conjunction with PDPH (Cat. No. 22301):
The method is outlined below:
1. Conjugate the target protein to the NHS ester (amine-reactive) group of Sulfo-SMCC. This will produce a maleimide-activated target protein via its primary amines (lysines and N-terminus).
2. Remove the excess sulfo-SMCC by desalting or dialysis.
3. React the maleimide-activated target protein to PDPH via sulfhydryls. (Lysines and N-terminus now are connected to hydrazide (amine) group with an extended linker)
4. Remove excess PDPH by desalting or dialysis.
5. React the hydrazide-activated target protein with carboxyl beads in the presence of EDC.
6. Use appropriate buffer to rinse off excess target protein from the carboxyl beads.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (3)

Citations & References
Abstract
Systemic administration of a nerve growth factor conjugate reverses age-related cognitive dysfunction and prevents cholinergic neuron atrophy.
Authors:Bäckman C, Rose GM, Hoffer BJ, Henry MA, Bartus RT, Friden P, Granholm AC
Journal:J Neurosci
PubMed ID:8757256
'Intraventricular administration of nerve growth factor (NGF) in rats has been shown to reduce age-related atrophy of central cholinergic neurons and the accompanying memory impairment. Intraventricular administration of NGF is necessary because NGF will not cross the blood-brain barrier (BBB). Here we have used a novel carrier system, consisting of ... More
Conjugation of folate via gelonin carbohydrate residues retains ribosomal-inactivating properties of the toxin and permits targeting to folate receptor positive cells.
Authors:Atkinson SF, Bettinger T, Seymour LW, Behr JP, Ward CM
Journal:J Biol Chem
PubMed ID:11359781
Conjugation of folate to proteins permits receptor-mediated endocytosis via the folate receptor (FR) and delivery of the conjugate into the cytoplasm of cells. Since many cancers up-regulate the FR it has enabled the targeting of toxins to tumor cells resulting in specific cell death. However, current conjugation methods rely on ... More
Site-specific modifications of light chain glycosylated antilymphoma (LL2) and anti-carcinoembryonic antigen (hImmu-14-N) antibody divalent f1agments.
Authors:Govindan SV, Goldenberg DM, Griffiths GL, Leung SO, Losman MJ, Hansen HJ
Journal:Cancer Res
PubMed ID:7493334
Site-specific introduction of metal-chelating groups into F(ab')2 fragments of an antilymphoma antibody (LL2) possessing a natural Asn-linked light chain carbohydrate and an anti-carcinoembryonic antigen antibody (hImmu-14-N) grafted with a light chain carbohydrate site is described. For this purpose, four yttrium- (and indium)-chelating agents were used, containing a primary amino group ... More