Pierce™ Sodium meta-Periodate
Thermo Scientific™
Pierce™ Sodium meta-Periodate
Thermo Scientific Pierce Sodium meta-Periodate is a gentle oxidizing agent that cleaves cis-diols in carbohydrate sugars to create amine-reactive aldehydes,Read more
Have Questions?
Catalog NumberQuantity
2050425 g
Catalog number 20504
Price (USD)
62.50
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
62.50
Each
Thermo Scientific Pierce Sodium meta-Periodate is a gentle oxidizing agent that cleaves cis-diols in carbohydrate sugars to create amine-reactive aldehydes, providing many uses relating to the study and detection of glycoproteins.
Features of Sodium meta-Periodate:
• Convert sugars in sialic acid and other glycosylation groups to reactive aldehydes
• Immobilize glycoproteins to a hydrazide-activated solid support
• Conjugate antibodies to glycoprotein enzymes, such as horseradish peroxidase
• Probe for cell-surface polysaccharides
• Detect carbohydrate-containing proteins using hydrazide-containing probes
Sodium meta-periodate converts cis-glycol groups in carbohydrates to amine-reactive aldehyde groups. Carbohydrate groups in glycoproteins are excellent sites for modification or crosslinking reactions because they allow the conjugation reaction to be directed away from amino acids in the polypeptide chain that could be critical for protein activity. Sodium meta-periodate cleaves bonds between adjacent carbon atoms that contain hydroxyl groups (cis-glycols), creating two aldehyde groups that are spontaneously reactive to amine- and hydrazide-activated labeling, immobilization supports and crosslinking reagents.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormPowder
Label TypeConjugating Groups
Label or DyeAldehyde Group
Product LinePierce
Quantity25 g
Product TypeSodium Meta-Periodate
Unit SizeEach
Contents & Storage
Upon receipt store at room temperature.
Frequently asked questions (FAQs)
What would be the best way to crosslink an antibody with horseradish peroxidase (‘HRP’ unmodified)?
A commonly used method for conjugating HRP to antibodies is by activation of the HRP using sodium meta-periodate (Cat no. 20504) to generate aldehydes. After the reaction, the HRP must be dialyzed to remove the meta-periodate; then the aldehydes on the HRP will react directly to the amine groups on the antibody. Alternatively, we offer activated HRP (Cat. No. 31488) as an alternative to avoid the periodate oxidation step to modify the HRP. 
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.