Sulfo-SDAD (Sulfo-NHS-SS-Diazirine) (sulfosuccinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate)
Thermo Scientific™
Sulfo-SDAD (Sulfo-NHS-SS-Diazirine) (sulfosuccinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate)
Thermo Scientific Pierce Sulfo-SDAD (Sulfo-NHS-SS-Diazirine) combines proven NHS-ester and diazirine-based photoreaction chemistries with conjugate amine-containing molecules with nearly any otherRead more
Have Questions?
Catalog NumberQuantity
2617550 mg
Catalog number 26175
Price (INR)
66,036.00
Each
Quantity:
50 mg
Request bulk or custom format
Price (INR)
66,036.00
Each
Thermo Scientific Pierce Sulfo-SDAD (Sulfo-NHS-SS-Diazirine) combines proven NHS-ester and diazirine-based photoreaction chemistries with conjugate amine-containing molecules with nearly any other functional group via long-wave UV-light activation. A 13.5Å spacer arm containing a cleavable disulfide bond separates the two photoreactive groups.
Features of Sulfo-SDAD (Sulfo-NHS-SS-Diazirine):
• Water soluble—solubility in aqueous solutions improved by a sulfonate group
• Heterobifunctional—NHS ester group reacts with primary amines at pH 7 to 9 to form covalent amide bonds; diazirine (azipentanoate) group reacts efficiently with any amino acid side chain or peptide backbone upon activation with long-wave UV light (330-370 nm)
• Controllable—two-step chemical crosslinking is activated using common laboratory UV lamps
• Easy to use—these crosslinkers are photo-stable under typical laboratory lighting conditions so there is no need to perform experiments in the dark
• Better than aryl azides—the diazirine photoreactive group has better photostability in normal light than phenyl azide groups of traditional photoreactive crosslinkers, yet the diazirine group is more efficiently activated by long-wave UV light
Succinimidyl-diazirine (SDA) reagents are a new class of crosslinkers that combine proven amine-reactive chemistry with an innovative and efficient diazirine-based photochemistry for conjugating amine-containing molecules to nearly any other functional group. The SDA crosslinkers include six compounds differing in spacer arm lengths, ability to cleave the crosslinked proteins, and presence or absence of a charged group for membrane permeability. Protein crosslinking is an important technique used to understand protein structure and to stabilize protein-protein interactions, and these SDA reagents extend the efficiency and range of interactions that can be explored by this approach.
Typical bifunctional amine- or sulfhydryl-reactive crosslinkers require specific amino acid groups (e.g., lysine or cysteine) at the correct distance on both proteins to capture protein interactions. The SDA crosslinkers circumvent this limitation by allowing specific labeling of one protein using the amine-reactive N-hydroxysuccinimide (NHS) ester followed by UV-activated crosslinking via the diazirine moiety to any amino acid side chain or peptide backbone of a second protein. Diazirine-based photocrosslinkers have better photostability than phenyl azide-based photocrosslinkers and are easily activated with long-wave UV light (330-370nm).
Multiple Diazirine Crosslinkers Available
The NHS-ester diazirine derivatives (SDA, LC-SDA and SDAD) lack a charged group and are membrane-permeable. This property makes them ideal for intracellular and intramembrane conjugations. By contrast, Sulfo-SDA, Sulfo-LC-SDA and Sulfo-SDAD contain negatively charged sulfate groups that improve their water solubility and reduce cell membrane permeability, enabling their use for extracellular protein crosslinking. SDAD and Sulfo-SDAD also have a disulfide bond within the spacer that can be cleaved with reducing agents.
• Short (3.9Å) and long (12.5Å) spacer arm varieties
• Membrane-permeable (NHS) and impermeable (Sulfo-NHS) varieties
• Non-cleavable and cleavable (disulfide spacer) varieties
Related Products
SDAD (NHS-SS-Diazirine)
Sulfo-SDA (Sulfo-NHS-Diazirine)
Sulfo-LC-SDA (Sulfo-NHS-LC-Diazirine)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
DescriptionSulfo-SDAD
FormSolid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)490.51
PEGylatedNo
Product LinePierce
Quantity50 mg
Reactive MoietyDiazirine, Sulfo-NHS Ester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length13.5 Å
Water SolubleYes
Chemical ReactivityAmine-Nonselective
CleavableBy Thiols
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C protected from light and moisture.
Frequently asked questions (FAQs)
What are the options for crosslinking proteins to nucleic acids, and what are the differences between them?
We offer the following crosslinkers for protein to nucleic acids: formaldehyde (Cat. No. 28906), SPB (Cat. No. 23013), SDAD (Cat. No. 26169), sulfo-SDAD (Cat. No. 26175), and sulfo-SBED (Cat. No. A39260).
Formaldehyde can be used for DNA-DNA crosslinking as well as DNA-protein crosslinking. While it is good for crosslinking proteins that are in direct contact with DNA, formaldehyde cannot link proteins that may be bound in a complex with other proteins but are not in direct contact with DNA. EGS or DSG can crosslink proteins to other proteins that are in direct contact with DNA and crosslinked to DNA by formaldehyde.
SPB, Sulfo-SDAD, and Sulfo-SBED are all photoreactive crosslinkers. SPB is an NHS-ester and psoralen heterobifunctional crosslinker that conjugates primary amines on proteins to DNA via photo-activated intercalation of psoralen to pyrimidine bases. The psoralen tricyclic planar ring system intercalates into double-stranded, and to a lesser extent, single-stranded DNA and RNA. The photoreactive psoralen group provides more selective covalent binding to nucleic acids than either phenyl azide or diazerine linkers, which makes SPB the best choice. The diazerine on sulfo-SDAD and the phenyl azide on sulfo-SBED are not selective for nucleic acids and can crosslink other biomolecules in the same besides any protein-DNA interactions. Sulfo-SBED involves biotin transfer rather than actual linking.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.