PageRuler™ Prestained NIR Protein Ladder
Thermo Scientific™

PageRuler™ Prestained NIR Protein Ladder

Thermo Scientific PageRuler Prestained NIR Protein Ladder is a mixture of 10 proteins (11 to 250 kDa) that are blue-stainedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
26635X48 x 250 μL
266352 x 250 mL
Catalog number 26635X4
Price (INR)
-
Quantity:
8 x 250 μL

Thermo Scientific PageRuler Prestained NIR Protein Ladder is a mixture of 10 proteins (11 to 250 kDa) that are blue-stained and fluor-labeled for near-IR fluorescent visualization and protein sizing following electrophoresis. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Product features

  • Fluorescent—detect using 670 nm red laser or 700 nm channels with fluorescence imagers
  • Reference band—55 kDa band has greater intensity for easy orientation

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on NIR fluorescent SDS-polyacrylamide gels and western blots
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodColorimetric, NIR Fluorescence (700 nm)
Molecular Weight (g/mol)250, 130, 95, 70, 55, 43, 34, 26, 15, 11 kDa
Quantity8 x 250 μL
Ready to LoadYes
ColorBlue
Number of Markers10
Product LinePageRuler
Product TypeProtein Ladder
Size Range11 to 250 kDa
System TypeSDS-PAGE
Unit SizeEach
Contents & Storage
Contents: eight vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C