Product Image
Thermo Scientific™
Immuno Clear Standard Modules
This product has been discontinued. Please consider Cat. No. 467679 as a replacement product.
Have Questions?
Catalog NumberSurface Treatment
6505TS
also known as 6505
Immulon 1 B
Catalog number 6505TS
also known as 6505
Price (INR)
-
Surface Treatment:
Immulon 1 B
This product has been discontinued. Please consider Cat. No. 467679 as a replacement product.
Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.
Features:
  • Several strip formats: Eight-well strip, 12-well strip, 16-well strip (two columns of eight wells)
  • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules that remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm
Choice of Surface:
  • Thermo Scientific™ PolySorp™, Immulon™ 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific™ Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific™ MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Modules are freestanding. Cat. No. 95029440 and 464394 are certified. Frames and modules are available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your customer service or sales representative for further details.
Specifications
Binding PropertyHydrophobic
Binding TypeAdsorption
CertifiedNo
FormatStandard
No. per Case100
No. per Pack100
Packaging100 Boxes
Product TypeMicroplate Strip
Target MoleculeLipids
Volume (Metric) Working280 μL
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Surface TreatmentImmulon 1 B
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 100
Frequently asked questions (FAQs)
What frames do you recommend for your Immuno Clear Standard Modules?
We recommend Frames and Accessories for Immuno Breakable Modules, BreakApart frame (Cat. No. 465404). This frame is designed to hold the Immuno Clear Standard Modules.