Evolution™ One/One Plus UV-Vis Spectrophotometers
Evolution™ One/One Plus UV-Vis Spectrophotometers
Evolution™ One/One Plus UV-Vis Spectrophotometers
Evolution™ One/One Plus UV-Vis Spectrophotometers
Thermo Scientific™

Evolution™ One/One Plus UV-Vis Spectrophotometers

Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeSpectral Bandwidth
840-370700Evolution OnePlus INTL USP/EP Standards Bundle1.0, 2.0 nm Variable
840-341400Evolution One UV-Vis Spec1 nm
840-341500Evolution One Plus UV-Vis Spec1.0, 2.0 nm Variable
840-370100Evolution One US Security Bundle1 nm
840-370500Evolution One INTL Security Bundle1 nm
840-370200Evolution One US USP/EP Standards Bundle1 nm
840-370600Evolution One INTL USP/EP Standards Bundle1 nm
840-370400Evolution OnePlus US Security Bundle1.0, 2.0 nm Variable
840-370800Evolution OnePlus INTL Security Bundle1.0, 2.0 nm Variable
840-370300Evolution OnePlus US USP/EP Standards Bundle1.0, 2.0 nm Variable
Catalog number 840-370700
Price (INR)
-
Request A Quote
Type:
Evolution OnePlus INTL USP/EP Standards Bundle
Spectral Bandwidth:
1.0, 2.0 nm Variable

For routine, complex research and advanced applications, including current compliance, the Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometer series brings together reliable, versatile hardware, a broad range of accessories, with easy-to-use, easy-to-learn Thermo Scientific™ Insight™ Pro Software.

Webinar: Identification of protein aggregation through UV-Vis spectroscopy

For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.

Optimized Engineering Delivers Ultimate Performance:
  • Double-beam geometry with fixed 1nm or variable spectral bandwidth options
  • Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories
Insight Pro Software—Easy-to-learn, easy-to-use
  • Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
  • Prevent costly sampling and measurement errors: Create dedicated workflows for complex assays and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
Accessories for all your Sampling Needs:
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
  • Easy software integration gives you complete control of your measurements

Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Evolution One/One Plus Security and Validator Bundles include:
  • Evolution One or One Plus UV-Vis Spectrometer
  • INSIGHT Pro with Security Software
  • Non-USP and EP Standards Set
Evolution One/One Plus USP/EP Standards Pharma Bundles include:
  • Evolution One or One Plus Spectrometer
  • INSIGHT Pro with Security Software
  • USP and EP UV Standards Set
  • USP and EP Visible Standards Set
Specifications
IncludesEvolution One Plus UV-Vis Spectrometer, INSIGHT Pro with Security Software, Validator Documentation A4 Ltr Paper Version, USP and EP UV Standards Set, USP and EP Visible Standards Set
Optical DesignAFBG Microcell optimized; AFBG Fiber optic optimized; AFBG Materials optimized
Product LineEvolution One Plus
Spectral Bandwidth1.0, 2.0 nm Variable
TypeEvolution OnePlus INTL USP/EP Standards Bundle
Unit SizeEach