Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL
Thermo Scientific™

Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL

Thermo Scientific Pierce High Capacity Endotoxin Removal Resin binds to reduce endotoxin levels in protein samples by ≥99% in 1Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8827525 Columns
882745 Columns
Catalog number 88275
Price (INR)
34,172.00
線上優惠
Ends: 31-Dec-2026
56,952.00
Save 22,780.00 (40%)
Each
Quantity:
25 Columns
Request bulk or custom format
Price (INR)
34,172.00
線上優惠
Ends: 31-Dec-2026
56,952.00
Save 22,780.00 (40%)
Each
Thermo Scientific Pierce High Capacity Endotoxin Removal Resin binds to reduce endotoxin levels in protein samples by ≥99% in 1 hour; with this spin cup format, protein recovery is ≥85%.

Features of High Capacity Endotoxin Removal Resin Spin Columns:

• High capacity—bind up to 2,000,000 EU/mL† to eliminate >99% of endotoxins
• Selective—recover ≥85% of your protein sample
• High performance—meets FDA guidelines by reducing final EU concentration to <5 EU/mL
• Fast—our new spin cup format enables endotoxin depletion within 1 hour
• Clean—single-use spin columns avoid cross contamination of samples
• Optimized—spin cups are optimized for different sample volumes

The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(ε-lysine), as an affinity ligand to selectively bind endotoxins. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers.

Requires:
Endotoxin-free water
Applications:

Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection

The presence of endotoxins in biologically derived products prepared for research, vaccination, or therapeutic use is a major concern due to the diverse and potentially harmful biological response to these molecules. Endotoxin contamination in proteins purified from bacterial cultures is due to residual lipopolysaccharides (LPS) derived from the outer cell membrane of gram-negative bacteria such as E. coli. Endotoxin-contaminated protein samples transfected into cells or injected into an animal host can initiate a strong immune response resulting in a Systemic Inflammatory Response Syndrome (SIRS) and/or sepsis. Traditional endotoxin removal resins that use polymixin B and sodium deoxycholate to clear endotoxins from a sample also have the negative side effects of neurotoxicity and acute renal tubular necrosis.

Pierce High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000-EU/mL binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/mL. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/mL. This spin cup format is a fast, single-use method to remove endotoxins from protein samples in as little as 1 hour. The spin cups use a batch format to bind and remove endotoxins while allowing for >85% protein recovery.

† One endotoxin unit/mL (EU/mL) equals approximately 0.1ng endotoxin/mL of solution.

More Product Data
• Removing endotoxins using a spin-column format
• Eliminate endotoxins from protein and antibody samples

Related Products
Pierce™ High Capacity Endotoxin Removal Resin
Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.25 mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)Up to 2,000,000 EU/mL
Column TypeAffinity, Cellulose Resin
FormatSpin Column
Formulation8mL-capacity centrifuge columns containing 0.5mL resin
Quantity25 Columns
Stationary PhaseProprietary Ligand
TargetEndotoxin Removal
FormLiquid
Product LinePierce
TypeSpin Column
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Citations & References (1)

Citations & References
Abstract
Unraveling function and diversity of bacterial lectins in the human microbiome.
Authors:Brady SF
Journal:Nature communications
PubMed ID:35661736