Axiom™ Propel Fast Reagents Kit, 96F
Applied Biosystems™

Axiom™ Propel Fast Reagents Kit, 96F

The Axiom Propel Fast Reagents Kit includes the reagents required to produce hybridization-ready targets for Axiom pre-designed and Axiom myDesignRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
9523714 x 96 arrays
9523728 x 96 arrays
Catalog number 952371
Price (INR)
800,275.00
Each
Number of Arrays:
4 x 96 arrays
Price (INR)
800,275.00
Each
The Axiom Propel Fast Reagents Kit includes the reagents required to produce hybridization-ready targets for Axiom pre-designed and Axiom myDesign arrays using the Multidrop Combi reagent dispensers. In addition, wash and stain reagents are included to process Axiom arrays using the GeneTitan MC instrument. Bulk reagent packaging allows the end user to process four or eight 96-format array plates with efficiency and reduced wastage.

The Axiom Propel Fast Reagent Kit can be used for genotyping of single nucleotide polymorphisms, copy number variants, and insertions/deletions on both Axiom Biobank Genotyping arrays and agrigenomics genotyping arrays.

Required products
Axiom Biobank Genotyping Arrays
Axiom Genotyping Solution for Agrigenomics
Axiom Propel Workflow for High-throughput Genotyping
Gene Titan Multi-Channel Instrument
GeneChip Command Console

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Genotyping
For Use With (Equipment)GeneTitan MC
IncludesReagent
No. of Samples4x96
Quantity4x96F
Reaction SpeedFast
TypeReagents Kit to Process 4X96F Array Plates
Format96-array Plate
Number of Arrays4 x 96 arrays
Unit SizeEach