Product Image
Applied Biosystems™
Axiom™ GeneTitan™ Scan Trays and Covers, bulk, 384HT format
Axiom GeneTitan Scan Trays and Covers allow for processing of ten Axiom 384HT-format array plates using the GeneTitan MC instrument.
Have Questions?
Catalog NumberQuantity
9523813840 samples
Catalog number 952381
Price (INR)
-
Quantity:
3840 samples
Axiom GeneTitan Scan Trays and Covers allow for processing of ten Axiom 384HT-format array plates using the GeneTitan MC instrument. Scan trays and covers are required along with GeneTitan barcoded stain trays/covers and hybridization trays to process arrays on the instrument. The bulk packaging is ideal for high throughput processing of arrays.
Related Links:
Specifications
Quantity3840 samples
TypeScan Tray and Cover
Format384 HT-array Plate
Number of Arrays3840 arrays
Unit SizeEach