Product Image
Applied Biosystems™
Axiom™ GeneTitan™ Barcoded Consumables, 1x384HT format
The Axiom GeneTitan Barcoded Consumables Kit includes all GeneTitan instrument barcoded consumables required to process one Axiom 384HT array plate.
Have Questions?
Catalog NumberQuantity
9523851 each
Catalog number 952385
Price (INR)
-
Quantity:
1 each
The Axiom GeneTitan Barcoded Consumables Kit includes all GeneTitan instrument barcoded consumables required to process one Axiom 384HT array plate. The barcodes alert the end user if the incorrect consumable has been loaded onto the GeneTitan MC instrument.
Related Links:
Specifications
Quantity1 each
TypeBarcoded Consumables Kit
Format384 HT-array Plate
Number of Arrays384 arrays
Unit SizeEach