TaqCheck™ SARS-CoV-2 Control
Applied Biosystems™

TaqCheck™ SARS-CoV-2 Control

The Applied Biosystems™ TaqCheck™ SARS-CoV-2 Control is a synthetic RNA positive control that contains target sequences for the SARS-CoV-2 S and N gene assays included in the TaqCheck™ SARS-CoV-2 Fast PCR Assay.
Have Questions?
Catalog NumberQuantity
9561273 x 10 μL
Catalog number 956127
Price (INR)
131,232.00
3 x 10 µL
Quantity:
3 x 10 μL
Price (INR)
131,232.00
3 x 10 µL
The Applied Biosystems TaqCheck SARS-CoV-2 Control is a synthetic RNA positive control that contains target sequences for the SARS-CoV-2 S and N gene assays included in the TaqCheck SARS-CoV-2 Fast PCR Assay.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™ 5 Real‐Time PCR Instrument, 384-well
Product LineTaqCheck
Quantity3 x 10 μL
Shipping ConditionDry Ice
SpeciesHuman
TargetSevere acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
For Use With (Application)Real Time PCR (qPCR)
PCR Method1-step RT-qPCR
Reaction SpeedFast
Unit Size3 x 10 µL