FreeStyle™ F17 Expression Medium
FreeStyle™ F17 Expression Medium
FreeStyle™ F17 Expression Medium
Gibco™

FreeStyle™ F17 Expression Medium

Gibco FreeStyle F17 Expression Medium is a chemically defined, protein-free, serum-free medium developed to support the growth and transfection ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A138350310 L
A13835011 L
A13835026 x 1 L
A138350420 L
Catalog number A1383503
Price (INR)
204,254.00
Each
Quantity:
10 L
Price (INR)
204,254.00
Each
Gibco FreeStyle F17 Expression Medium is a chemically defined, protein-free, serum-free medium developed to support the growth and transfection of HEK293 cells in suspension. The addition of 4–8 mM L-glutamine or GlutaMAX supplement is required before use.

Features:
• Little or no required adaptation
• Supports suspension growth and transfection of HEK293 cells
• Chemically defined, protein-free, serum-free formulation
• Compatible with multiple transient production systems
• Scalability in spinner flasks and bioreactors

Little or no required adaptation
293-F, 293-H, and FreeStyle 293-F cells in suspension can be cultured directly into FreeStyle F17 Expression Medium without adaptation. Cells adapted to other commercially available serum-free media can be subcultured directly into FreeStyle F17 medium, usually without any further adaptation. Cells usually require adaptation from serum-containing formulations.

Growth and transfection of HEK293 cells
FreeStyle F17 Expression Medium supports 293-F cells at a peak density of 6–7 x 106 viable cells per mL. Cells cultured in FreeStyle F17 medium can be transfected using a wide variety of transfection reagents, including lipid-based transfection reagents, such as 293fectin and Freestyle MAX Reagent.

Scalability in spinner flasks and bioreactors
Production using FreeStyle F17 Expression Medium can be scaled up in spinner flasks or bioreactors. The appropriate spinner or impeller speed and seeding density should be optimized for each system. Depending on the impeller design and speed, it may be necessary to supplement FreeStyle F17 medium with additional Pluronic F-68 to avoid sheer stress in the culture.

cGMP manufacturing and quality system
FreeStyle F17 Expression Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Design your own medium specific to your needs
Whether it’s a bench-level experiment or large-scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco cGMP Media Custom Services you can put the manufacturing power and years of technical experience to work for you.
For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHEK293
Concentration1 X
Culture EnvironmentCO2
Endotoxin Level< 1 EU/mL
For Use With (Application)Protein expression, viral vector production
Product LineFreestyle
Product TypeExpression Medium
Purity or Quality GradeQualified for Further Manufacturing
Quantity10 L
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeSuspension Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile
Without AdditivesNo Glutamine
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How long can I keep culturing FreeStyle cells after transfection?

We typically keep the culture going for 7-10 days posttransfection. After that, protein expression may drop.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between FreeStyle 293 Expression Medium and FreeStyle F17 Expression Medium?

FreeStyle F17 Expression Medium was designed from a different base formulation than FreeStyle 293 Expression Medium. FreeStyle F17 medium shows equal or better performance than the FreeStye 293 medium for certain cell lines and proteins. It is capable of supporting the growth of 293 and CHO cells and offers a wider variety and selection of transient cell culture media options.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.