Product Image
Invitrogen™

Silencer™ Human GPCR siRNA Library

G-protein–coupled receptors (GPCRs) are key components of many signal transduction pathways and also represent an important class of druggable genes.Read more
Have Questions?
Catalog NumberQuantity
A300821 Set
Catalog number A30082
Price (INR)
463,307.00
Online offer
Ends: 31-Dec-2026
772,177.00
Save 308,870.00 (40%)
Each
Quantity:
1 Set
Price (INR)
463,307.00
Online offer
Ends: 31-Dec-2026
772,177.00
Save 308,870.00 (40%)
Each
G-protein–coupled receptors (GPCRs) are key components of many signal transduction pathways and also represent an important class of druggable genes. The Silencer™ Human GPCR siRNA Library targets 1,134 Ambion™ Silencer siRNAs representing 378 non-olfactory human GPCRs. This library is designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC). Utilizing Ambion Silencer technology built on stringent bioinformatics to enable high target specificity and siRNA potency, the Silencer Human GPCR siRNA Library allows accurate yet cost-effective RNAi functional screening against known GPCRs.

• Comprehensive—Expedite functional gene discovery with pre-defined Silencer Human GPCR siRNA Library
• Reliable—Increase target screening accuracy with three individual siRNAs per gene target
• Effective—Reduce off target impacts at low concentrations with proven Ambion Silencer siRNA design
• Convenient—Reduce data de-convolution and accelerate target validation with our arrayed format
• Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats

Why use the Silencer Human GPCR siRNA Library?
Together with careful bioinformatics selection of gene targets, Silencer siRNAs are optimized for design and high quality synthesis with guaranteed silencing (learn more about the guarantee). The Silencer Human GPCR siRNA Library contains our pre-designed and when available, validated siRNAs generated with maximum potency and specificity in mind so you can focus on data quality (learn more about the design). All Silencer libraries are selected against the latest available databases and resources for up-to-date content. Full siRNA sequence information, as well as the exon targeted and each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID, plus results from any validation experiments performed using the included siRNAs is provided. Please contact us for the complete list of targets.

Silencer siRNA libraries allow you to screen efficiently and maximize data output
Ambion Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps to ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human GPCR siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).

Flexibility to meet your workflow needs
Thermo Fisher Scientific offers comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.com.

Specifications
For Use With (Application)RNAi
Product LineSilencer
Product TypesiRNA Library
PurityStandard
Quantity1 Set
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Format96-well Plate
RNAi TypesiRNA
SpeciesHuman
Unit SizeEach
Contents & Storage
1134 total siRNAs targeting 378 genes plated onto 96-well plates (15 plates)

Store at or below -20°C in a non-frost-free freezer.

Frequently asked questions (FAQs)

Which Silencer siRNA libraries do you offer?

Our Silencer siRNA library offerings can be found here (https://www.thermofisher.com/us/en/home/life-science/rnai/synthetic-rnai-analysis/sirna-libraries/silencer-sirna-libraries.html).

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.

Which Silencer Select siRNA libraries do you offer?

Our Silencer Select siRNA library offerings can be found here (https://www.thermofisher.com/us/en/home/life-science/rnai/synthetic-rnai-analysis/sirna-libraries/silencer-select-sirna-libraries.html).

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.