Alexa Fluor™ 405 Cadaverine
Invitrogen™

Alexa Fluor™ 405 Cadaverine

Alexa Fluor™ 405 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via aRead more
Have Questions?
Catalog NumberQuantity
A30675
also known as A-30675
1 mg
Catalog number A30675
also known as A-30675
Price (INR)
48,509.00
Each
Quantity:
1 mg
Price (INR)
48,509.00
Each
Alexa Fluor™ 405 Cadaverine is useful as a polar tracer and as a reactive dye for labeling proteins via a carboxylic acid moiety. Alexa Fluor™ 405 is a bright, violet dye with excitation ideally suited to the 405 nm laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 405 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 405 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

Detailed information about this AlexaFluor™ cadaverine:

• Fluorophore label : Alexa Fluor™ 405 dye
• Reactive group: cadaverine
• Reactivity: carboxylic acids, aldehydes, and ketones (and glutamine residues through an enzyme-catalyzed transamidation reaction)
• Ex/Em of the conjugate: 399/422 nm
• Extinction coefficient: 29,000 cm-1M-1
• Spectrally similar dyes: Cascade Blue
• Molecular weight: 666.58

Cell Tracking and Tracing Applications
Alexa Fluor™ cadaverines make excellent fluorescent polar tracers because they are bright, small, and water soluble. Since they contain an aldehyde-fixable functional group, they can be fixed in cells by treatment with formaldehyde or glutaraldehyde. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent. Learn more about cell tracking and tracing.

Protein Labeling Applications
Alexa Fluor™ cadaverines can be used as reactive molecules for adding a fluorescent label to carboxylic acids using a coupling agent such as a carbodiimide; they do not spontaneously react with carboxylic acids in solution. They do, however, react spontaneously with the common amine-reactive functional groups, including succinimidyl esters and isothiocyanates. The amine-containing Alexa Fluor™ cadaverines can also be used to label glutamine residues in some proteins and peptides via an enzyme-catalyzed transamidation reaction.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products
DMSO (dimethylsulfoxide) (D12345)
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityCarboxylic Acid, Ketone, Aldehyde
Emission422 nm
Excitation399 nm
Label or DyeAlexa Fluor™ 405
Product TypeCadaverine
Quantity1 mg
Reactive MoietyAmine, Cadaverine
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (2)

Citations & References
Abstract
Whole-cell 3D STORM reveals interactions between cellular structures with nanometer-scale resolution.
Authors:Huang B, Jones SA, Brandenburg B, Zhuang X,
Journal:Nat Methods
PubMed ID:19029906
The ability to directly visualize nanoscopic cellular structures and their spatial relationship in all three dimensions will greatly enhance our understanding of molecular processes in cells. Here we demonstrated multicolor three-dimensional (3D) stochastic optical reconstruction microscopy (STORM) as a tool to quantitatively probe cellular structures and their interactions. To facilitate ... More
Cascade blue derivatives: water soluble, reactive, blue emission dyes evaluated as fluorescent labels and tracers.
Authors:Whitaker JE, Haugland RP, Moore PL, Hewitt PC, Reese M, Haugland RP
Journal:Anal Biochem
PubMed ID:1724118
Fluorescent dyes based on the pyrenyloxytrisulfonic acid (Cascade Blue) structure were prepared and evaluated. The dyes contain functional groups that react with amines, thiols, acids, aldehydes, and ketones, forming covalently bonded, fluorescent derivatives of molecules with broad biological interest. Reactive groups in the Cascade Blue dyes include carboxylic acids and ... More