Torrent Storage™ NAS Device
Ion Torrent™

Torrent Storage™ NAS Device

The Torrent Storage™ NAS Device provides network-attached storage (NAS) for Ion Torrent™ sequencing runs and increases available storage space onRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A321991 Server, 9 x 8 TB Drives
A321981 Server, 9 x 4 TB Drives
A322001 Server, 18 x 8 TB Drives
Catalog number A32199
Price (INR)
-
Quantity:
1 Server, 9 x 8 TB Drives
The Torrent Storage™ NAS Device provides network-attached storage (NAS) for Ion Torrent™ sequencing runs and increases available storage space on your Torrent Server. The NAS Device connects to the Torrent Server directly or through your local network. Torrent Storage NAS devices are installed by one of our field service engineers.

Save data in your lab or data center
Share storage with multiple Ion S5™ instruments and Torrent Servers
Transfer data over direct network or LAN
Access stored data over your network
Use reliable storage with disk failure tolerance
Expand storage as needed*

Onsite installation and configuration
After the NAS Device is ordered, it is installed locally by a field service engineer to ensure proper configuration.

Direct file export from Torrent Suite Software
Files generated from an Ion instrument are saved on the Torrent Server and can be transferred directly to the NAS Device via the Torrent Suite™ software interface. After transfer of the data, the files are available for re-analysis or archiving.

Torrent Storage NAS 110T device capacity
File typeIon 318™ ChipIon 520™ ChipIon 530™ ChipIon 540™ Chip
WELLS and BAM785055001800450
BAM files only220002200084002100

Note: These numbers are approximate based on 200 bp runs.

Services and support
Service and support plans—comprehensive plans to maximize instrument performance and ensure availability of critical systems
Training—courses for experimental design, data analysis, troubleshooting, applications, and instrument operation
Bioinformatics—tailored support for NGS informatics execution, software use, data analysis, and downstream applications
DataSafe Solution—storage and back-up strategy, design, hardware, and support for all lab data

*Storage expansion is limited.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion Proton™ System
Product TypeNAS Device
Quantity1 Server, 9 x 8 TB Drives
Shipping ConditionRoom Temperature
Product LineTorrent Storage
Software TypeTorrent Suite Software
Unit SizeEach
Contents & Storage
1 Dell T630 Server with 9 x 8 TB drives

Frequently asked questions (FAQs)

How do I set up a network file sharing (NFS) to archive or backup data from Torrent Suite Software?

Settings depend on the file server that you have acquired. The destination folder must have read and write permissions. If you are using network file sharing (NFS), which we recommend, the “no_root_squash” option must be turned on.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How can I backup my data in Torrent Suite Software?

We provide a Torrent NAS (Network Attached Server - https://www.thermofisher.com/order/catalog/product/A32198. We can also provide other backup devices tailored to your needs - see more at https://www.thermofisher.com/order/catalog/product/A27662.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.