Ion S5™ Precision ID Chef & Sequencing Kit (2 runs per initialization)
Applied Biosystems™

Ion S5™ Precision ID Chef & Sequencing Kit (2 runs per initialization)

The Ion S5 Precision ID Chef & Sequencing Kit enables robust and automated template preparation and sequencing of Precision ID libraries using the Ion Chef System and an Ion GeneStudio S5 sequencing system (four initializations and eight sequencing runs).
Have Questions?
Catalog NumberQuantity
A332088 reactions
Catalog number A33208
Price (INR)
668,288.00
Each
Quantity:
8 reactions
Price (INR)
668,288.00
Each

The Ion S5 Precision ID Chef & Sequencing Kit enables robust and automated template preparation and sequencing of Precision ID libraries using the Ion Chef System and an Ion GeneStudio S5 sequencing system (four initializations and eight sequencing runs). Using this kit with the Ion S5 workflow provides template preparation and chip loading reagents for libraries generated with commercially available Precision ID panels (GlobalFiler NGS STR Panel v2, Identity Panel, Ancestry Panel, mtDNA Whole Genome Panel, mtDNA Control Region Panel, or custom Ion AmpliSeq SNP panel). For eight initializations and eight sequencing runs, see Cat. No. A35850.

The Ion S5 Precision ID Chef & Sequencing Kit for the Ion Chef and Ion GeneStudio S5 workflow:

  • Provides template preparation and chip loading reagents for Precision ID libraries
  • Simplifies your testing with ready-to-use, disposable reagent cartridges for sequencing
  • Delivers fast and simple sequencing instrument set-up, with less than 15 minutes hands-on time
  • Leverages our latest advancements in data quality and accuracy
  • Maximizes flexibility through compatibility with both the Ion 520 and Ion 530 chips

The Ion GeneStudio S5 sequencers, along with the Ion Chef System, provide reproducible sequencing workflows with minimal hands-on time. A single Ion 530 Chip accommodates 32 libraries made with the GlobalFIler NGS STR Panel v2.

Specifications
Product TypePrecision ID Chef and Sequencing Kit
Quantity8 reactions
Shipping ConditionApproved for shipment on Wet or Dry Ice
Product LineIon S5
Unit SizeEach
Contents & Storage
• Ion S5 Precision ID Chef solutions, store at room temperature
• Ion S5 Precision ID Chef supplies, store at room temperature
• Ion S5 Precision ID Chef reagents, store at -5°C to -30°C
• Ion S5 Precision ID sequencing solutions, store at room temperature
• Ion S5 Precision ID sequencing reagents, store at -5°C to -30°C

Frequently asked questions (FAQs)

What are the contents of Ion S5 Precision ID Sequencing Solutions (Part No. A33211)?

The box contains the following:

- Precision ID Wash Solution, 1.5 L, qty. 4
- Precision ID Cleaning Solution, 250mL, qty. 1

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is the storage condition for Ion S5 Precision ID Sequencing Solutions (Part No. A33211)?

We recommend storing at 15 to 30 degrees C.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is the storage condition for Ion S5 Precision ID Chef Reagents (Part No. A32882)?

We recommend storing at -30 to -10 degrees C.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What are the contents of Ion S5 Precision ID Chef Reagents (Part No. A32882)?

The box contains Ion S5 Precision ID Chef Reagents cartridges, qty. 4.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What is the storage condition for Ion S5 Precision ID Chef Solutions (Part No. A33210)?

We recommend storing at 15 to 30 degrees C.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.