Ion S5™ Precision ID Chef & Sequencing Kit (2 runs per initialization) - FAQs
Ion S5™ Precision ID Chef & Sequencing Kit (2 runs per initialization) - FAQs
View additional product information for Ion S5™ Precision ID Chef & Sequencing Kit (2 runs per initialization) - FAQs (A33208)
15 product FAQs found
What are the contents of Ion S5 Precision ID Sequencing Solutions (Part No. A33211)?
The box contains the following:
- Precision ID Wash Solution, 1.5 L, qty. 4
- Precision ID Cleaning Solution, 250mL, qty. 1
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for Ion S5 Precision ID Sequencing Solutions (Part No. A33211)?
We recommend storing at 15 to 30 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for Ion S5 Precision ID Chef Reagents (Part No. A32882)?
We recommend storing at -30 to -10 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the contents of Ion S5 Precision ID Chef Reagents (Part No. A32882)?
The box contains Ion S5 Precision ID Chef Reagents cartridges, qty. 4.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for Ion S5 Precision ID Chef Solutions (Part No. A33210)?
We recommend storing at 15 to 30 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the contents of Ion S5 Precision ID Chef Solutions (Part No. A33210)?
The box contains Ion S5 Precision ID Chef Solutions cartridges, qty. 4.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the contents of Ion S5 Precision ID Sequencing Reagents (Part No. A32883)?
The box contains Ion S5 Precision ID Sequencing Reagents cartridges, qty. 4.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for Ion S5 Precision ID Sequencing Reagents (Part No. A32883)?
We recommend storing at -30 to -10 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the storage condition for Ion S5 Precision ID Chef Supplies (Part No. A33209)?
We recommend storing at is 15 to 30 degrees C.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the contents of Ion S5 Precision ID Chef Supplies (Part No. A33209)?
The box contains the following:
- Chip adapter, qty. 2
- Enrichment Cartridge v2, qty. 1
- Tip Cartridge v2, qty. 1
- PCR Plate and Frame Seal v2, 1 each
- Recovery Station Disposable Lid v2, qty. 2
- Recovery Tubes, qty. 12
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Which Precision ID panels can be used with the Ion S5 Precision ID Chef & Sequencing Kit (Cat. Nos. A35850, A33208)?
The kits are compatible with the following commercially available Precision ID panels: GlobalFiler NGS STR panel v2, Identity panel, Ancestry panel, mtDNA Whole Genome panel, mtDNA Control Region panel, and Precision ID SNP panel.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the storage conditions for the Ion S5 Precision ID Chef & Sequencing Kit (Cat. Nos. A35850, A33208)?
Here are the storage conditions for the individual kit components:
- Ion S5 Precision ID Chef Solutions, store at room temperature
- Ion S5 Precision ID Chef Supplies, store at room temperature
- Ion S5 Precision ID Chef Reagents, store at -5 to -30 degrees C
- Ion S5 Precision ID Sequencing Solutions, store at room temperature
- Ion S5 Precision ID Sequencing Reagents, store at -5 to -30 degrees C
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What is the shipping condition for the Ion S5 Precision ID Chef & Sequencing Kit (Cat. Nos. A35850, A33208)?
The kits are shipped on wet or dry ice.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many runs can be performed with Ion S5 Precision ID Chef & Sequencing Kit (2 runs per initialization)?
The kit provides components for 8 initializations (8 runs at 2 runs per initialization) on the Ion S5 system.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the components of the Ion S5 Precision ID Chef & Sequencing Kit (2 runs per initialization)?
The kit provides components for 4 Ion S5 Sequencer initializations (for 8 runs at 2 runs per initialization). Here are the kit components:
- Ion S5 Precision ID Chef Supplies, Part No. A33209, qty. 4
- Ion S5 Precision ID Chef Reagents, Part No. A32882, qty. 1
- Ion S5 Precision ID Chef Solutions, Part No. A33210, qty. 1
- Ion S5 Precision ID Sequencing Reagents, Part No. A32883, qty. 1
- Ion S5 Precision ID Sequencing Solutions, Part No. A33211, qty. 1
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.