TaqMan™ Advanced miRNA Human A Card
Applied Biosystems™

TaqMan™ Advanced miRNA Human A Card

The Applied Biosystems TaqMan Advanced miRNA Human A Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in aRead more
Have Questions?
Catalog NumberQuantity
A34714384 reactions
Catalog number A34714
Price (INR)
64,228.00
Each
Quantity:
384 reactions
Price (INR)
64,228.00
Each
The Applied Biosystems TaqMan Advanced miRNA Human A Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format. This product contains one micro-fluidic card, enabling accurate quantitation of 384 unique microRNAs in as little as five hours. Four TaqMan Advanced miRNA Assay endogenous controls are included in the card to aid in data normalization. Details about the included assays are available in the Target List file. The assays can detect and quantify the mature form of the miRNA from 1–10 ng of total RNA from tissue.

Note: The TaqMan Advanced miRNA cDNA Synthesis Kit is required for preparation of the cDNA template that is used with the TaqMan Advanced miRNA assays.

The kit enables the analysis of:
• Multiple miRNAs from a single amplified sample
• Samples that are limited in quantity, including serum, plasma, or other biological fluids

Key features include:
Universal reverse transcription (RT)—one RT step for all TaqMan Advanced miRNA assays
Sensitive—detect as few as 60 copies of input miRNA
Specific—detect only mature miRNA and distinguish between highly homologous miRNAs
Small sample input—detect and quantify mature miRNA from as little as 1–10 ng of total RNA or 2 μL of plasma or serum
Versatile—compatible with tissue and biofluids, including serum and plasma

TaqMan Advanced miRNA cards provide all the advantages of TaqMan Advanced miRNA assays in a convenient, pre-configured microfluidic card, minimizing the experimental variability and effort required to run 384 TaqMan Advanced miRNA assays in parallel. With the TaqMan Advanced miRNA cDNA Synthesis Kit, TaqMan Advanced miRNA cards enable a comprehensive expression profile consistent with miRBase v21, providing the ideal miRNA profiling solution.

TaqMan Advanced miRNA cards are compatible with the following Applied Biosystems qPCR instruments: Viia7, QuantStudio 7 Flex, and QuantStudio 12k Flex.

Recommended master mix: TaqMan Fast Advanced Master Mix

TaqMan Advanced miRNA cards require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal RT chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.

These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well microfluidic card
Product LineTaqMan
Product TypeAdvanced miRNA Human A Card
Quantity384 reactions
Research CategorymiRNA
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
SpeciesHuman
Volume2 μL
Detection MethodPrimer-probe
For Use With (Application)miRNA analysis
GC-Rich PCR PerformanceHigh
Label or Dye2 Color
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
One 384-well array

The TaqMan miRNA card is shipped at ambient temperature. Upon arrival, store at 2–8°C and keep protected from light.

Frequently asked questions (FAQs)

I am not seeing good amplification with my TaqMan Advanced miRNA array cards. What is wrong?

The TaqMan Advanced miRNA array cards allow one to profile over 300 different miRNA targets on one card. Due to the large number of targets and the nature of your sample, it is possible that some miRNA on the card will not be expressed on your sample, or expressed at very low levels. This is one reason why you will not see good amplification plots for every spot on the array card. Another possibility is the sample quality or input. Make sure to check the concentration and quality of the RNA sample before reverse transcription. Check the Ct values for your endogenous control assays on the card, and see if it is in the expected range for your typical samples.